Subtotal = $0.00 
Items = 0 
View Cart
United States
1-800-323-4340 or 
Welcome,
log in or register
Price Range:
Custom Ordering Solutions
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here. Call 1-800-323-4340 Option 9 ext. 6334
 

Tris Hydrochloride

Tris Hydrochloride

  • Assay - ≥99%
  • IR - Conforms to standard
  • Melting Point - 152°±2°C
  • Absorbance of a 1M Solution at 280nm - ≤0.05
  • Calcium - ≤10ppm
  • Copper - ≤1ppm
  • Iron - ≤2ppm
  • Magnesium - ≤10ppm
  • Lead - ≤5ppm
  • Arsenic - ≤10ppm
  • Barium - ≤1ppm
  • Manganese - ≤1ppm
  • Electrophoresis - To pass test
  • DNase - Not detected
  • RNase - Not detected
  • Protease - Not detected
 Product Detail
Sort
 Availability
 Price
Representative photo only Tris Hydrochloride Small White Flakes Molecular Biology 1kg

Tris Hydrochloride (Small White Flakes/Molecular Biology) (1kg)

Product Type Chemical Bio-Reagents,Purity grade Molecular Biology,Form Flakes
EW-88057-53
In stock.
$206.00 USD/EACH

Qty:

Representative photo only Tris Hydrochloride Small White Flakes Molecular Biology 500g

Tris Hydrochloride (Small White Flakes/Molecular Biology) (500g)

Product Type Chemical Bio-Reagents,Purity grade Molecular Biology,Form Flakes
EW-88057-54
In stock.
$166.00 USD/EACH

Qty:

2 results (showing 1 - 2 )
Per Page 

View products by: Title | Brand | Category | Price | Popularity | Best Sellers
View product categories by: Popularity | Our Choices | All-Around Favorites | Title | Product Category | Recommended
View popular items by: Popularity | Favorites | Title | Price | Part Number | Category
View Popular Categories by: Popularity | Our Choices | All-Around Favorites | Product Category | Recommended | Title