log in or register
Price Range:
Custom Ordering Solutions
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here. Call 1-800-323-4340 Option 9 ext. 6334
Tubing Weights
Tubing Weights
Tubing weights in PTFE and stainless steel. PTFE weights keep tubing in place during dispensing and fluid transfer applications; stainless steel flow-through weight prevents tubing from curling or floating and is ideal for groundwater and well sampling.
 Product Detail
Sort
 Availability
 Price
EW-78226-82 PTFE Flow Through Tubing Weight for 5/16 to 7/16" ID Tubing.
EW-78226-82
In stock.
$34.50 USD/EACH
Qty:
See all Tubing Weights
EW-78226-81 PTFE Flow Through Tubing Weight for 3/16 to 1/4" ID Tubing.
EW-78226-81
In stock.
$24.00 USD/EACH
Qty:
See all Tubing Weights
Representative photo only Stainless steel flow through tubing weight
Stainless steel flow-through tubing weight.
Product Type Tubing Weights
EW-07570-04
In stock.
$72.00 USD/EACH
Qty:
See all Tubing Weights
EW-77310-03 PTFE flow-through tubing weights, set of two.
PTFE flow-through tubing weights, set of two.
Product Type Tubing Weights
EW-77310-03
In stock.
$49.90 USD/SET of 1
Qty:
See all Tubing Weights
4 results (showing 1 - 4 )
Per Page 
View products by: Title | Brand | Category | Price | Popularity | Best Sellers
View product categories by: Popularity | Our Choices | All-Around Favorites | Title | Product Category | Recommended
View popular items by: Popularity | Favorites | Title | Price | Part Number | Category
View Popular Categories by: Popularity | Our Choices | All-Around Favorites | Product Category | Recommended | Title