log in or register
Category:
Brand:
Product Type:
Availability:
CE Compliance:
Yes (85)
Max flow rate (GPM):
Max flow rate (LPM):
7.6 (1)
15 (1)
2,600 pages
of answers
FREE
2013/14 Cole-Parmer
Sourcebook
88 pages of flow meters and hundreds of pages of flow accessories including pumps, fittings, tubing and process control.
Request your copy today.
Custom Ordering Solutions
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here. Call 1-800-323-4340 Option 9 ext. 6334
Turbine Flowmeters
Turbine Flowmeters
Cole Parmer offers an extensive range of turbine flow meters for both liquid and gas applications. There are many types of turbine flow meters, some offer low cost water meter solutions with accuracy typically around 3% full scale, while other turbine flowmeters offer high accuracy of 1% or better.
Cole-Parmer's turbine flow meter selection encompasses a wide range of materials including low cost nylon and aluminium bodies to specialty options such as PVDF or stainless steel for more aggressive, higher temperature applications. Many of our turbine flowmeters come with a wide range of digital displays and controllers including options for PC, PLC control or batch dispensing operation.
How a Turbine Flowmeter Works
turbine flowmeter This mechanism includes a bladed rotor that is positioned along the centerline of the flow stream. The rotating component is designed to provide a pulse when passing either a magnetic or optical sensor. The frequency of the pulses is proportional to the velocity of the fluid. Some designs offer high levels of accuracy and can often handle slightly higher viscosity fluids than basic propeller-type designs. Some turbine designs meet sanitary guidelines (where stipulated by the manufacturer).
Typical Applications for turbine flow meters
  • Pilot plants
  • Research and development facilities
  • Cooling water monitoring
  • Inventory control metering applications
  • Test stands
  • Water consumption
  • Makeup water
Advantages for turbine flow meters
  • High accuracy and good repeatability
  • Fast response timetypically a few milliseconds
  • Wide flow range options for both liquids and gases
  • Good flexibility with associated digital readout devices including flow control options for both PC and PLC's
Disadvantages for turbine flow meters
  • Require upstream straightener and filter for best results
  • May over-read in pulsating flows
  • Turbine flow meters for liquid can require regular re-calibration due to bearing wear
  • Particulates can cause line blockage
To find the right flow meter for your application use Cole-Parmer's on-line flow meter quick search. Alternatively contact our Application Specialists for free technical support and let us bring over 50 years of flow control experience to help solve your application needs.
Still have questions?
Contact Our Experts
for FREE product
application selection
and support.
or call 1-800-323-4340
 Product Detail
Sort
 Availability
 Price
Representative photo only Battery powered turbine flowmeter totalizer 3 4 NPT F 2 to 20 gpm 7 6 to 75 7 LPM 316 SS
Battery-powered turbine flowmeter/totalizer, 3/4" NPT(F), 2 to 20 gpm/7.6 to 75.7 LPM, 316 SS
Product Type Turbine Flowmeters,Description Turbine Flowmeter,Max operating temperature º F 140
EW-05609-09
In stock.
$716.00 USD/EACH
Qty:
Representative photo only A109GMN025NA1 Flmtr 3 3GPM Digital Flowmeter W 1 4inNYLON Turbine Housing 2TOTALS 2CALS And Flowrate
A109GMN025NA1:Flmtr .3/3GPM Digital Flowmeter W/ 1/4inNYLON Turbine Housing 2TOTALS 2CALS And Flowrate
Product Type Turbine Flowmeters,Description Turbine Flowmeter,Max operating temperature º F 140
EW-05610-01
In stock.
$483.00 USD/EACH
Qty:
Representative photo only Battery powered turbine flowmeter totalizer 1 1 2 Tri Clover 5 to 50 gpm 18 9 to 190 LPM 316 SS
Battery-powered turbine flowmeter/totalizer, 1 1/2" Tri-Clover, 5 to 50 gpm/18.9 to 190 LPM, 316 SS
Product Type Turbine Flowmeters,Description Turbine Flowmeter,Max operating temperature º F 140
EW-05609-17
In stock.
$849.00 USD/EACH
Qty:
Representative photo only Battery powered turbine flowmeter totalizer 1 NPT F 5 to 50 gpm 18 9 to 190 LPM 316 SS
Battery-powered turbine flowmeter/totalizer, 1" NPT(F), 5 to 50 gpm/18.9 to 190 LPM, 316 SS
Product Type Turbine Flowmeters,Description Turbine Flowmeter,Max operating temperature º F 140
EW-05609-15
In stock.
$730.00 USD/EACH
Qty:
Representative photo only Battery powered turbine flowmeter totalizer 2 NPT F 20 to 200 gpm 76 to 760 LPM 316 SS
Battery-powered turbine flowmeter/totalizer, 2" NPT(F), 20 to 200 gpm/76 to 760 LPM, 316 SS
Product Type Turbine Flowmeters,Description Turbine Flowmeter,Max operating temperature º F 140
EW-05609-31
Usually ships in 10 days.
$1,274.00 USD/EACH
Qty:
Representative photo only Battery powered turbine flowmeter totalizer 1 2 NPT F 1 to 10 gpm 3 8 to 37 9 LPM 316 SS
Battery-powered turbine flowmeter/totalizer, 1/2" NPT(F), 1 to 10 gpm/3.8 to 37.9 LPM, 316 SS
Product Type Turbine Flowmeters,Description Turbine Flowmeter,Max operating temperature º F 140
EW-05609-03
In stock.
$699.00 USD/EACH
Qty:
Representative photo only A109GMN100NA1 Flmtr 3 50GPM Digital Flowmeter W 1inNYLON Turbine Housing 2TOTALS 2CALS And Flowrate
A109GMN100NA1:Flmtr 3/50GPM Digital Flowmeter W/ 1inNYLON Turbine Housing 2TOTALS 2CALS And Flowrate
Product Type Turbine Flowmeters,Description Turbine Flowmeter,Max operating temperature º F 140
EW-05610-04
Usually ships in 9 days or sooner.
$485.00 USD/EACH
Qty:
Representative photo only PVDF Infrared Flow Sensor 0 13 to 3 96 GPM 7 16 barb connection
PVDF Infrared Flow Sensor;
0.13 to 3.96 GPM, 7/16" barb connection
Product Type Turbine Flowmeters,Description Turbine Flowmeter,Max operating temperature º F 185
EW-32250-22
Usually ships in 6 days.
$508.00 USD/EACH
Qty:
Representative photo only Battery powered turbine flowmeter totalizer 3 4 Tri Clover 1 to 10 gpm 3 8 to 37 9 LPM 316 SS
Battery-powered turbine flowmeter/totalizer, 3/4" Tri-Clover, 1 to 10 gpm/3.8 to 37.9 LPM, 316 SS
Product Type Turbine Flowmeters,Description Turbine Flowmeter,Max operating temperature º F 140
EW-05609-05
In stock.
$822.00 USD/EACH
Qty:
EW-32610-44 Sample image only.  Check individual details.
Precision Turbine Flowmeter, 1.6 to 16 GPM, 3/4" NPT(M), with display and 4-20 mA output
Product Type Turbine Flowmeters,Description Turbine Flowmeter,Max flow rate GPM 1.6 to 16.0
EW-32610-44
Usually ships in 15 days.
$1,470.00 USD/EACH
Qty:
Representative photo only Battery powered turbine flowmeter totalizer 2 NPT F 20 to 200 gpm 76 to 760 LPM Aluminum
Battery-powered turbine flowmeter/totalizer, 2" NPT(F), 20 to 200 gpm/76 to 760 LPM, Aluminum
Product Type Turbine Flowmeters,Description Turbine Flowmeter,Max operating temperature º F 140
EW-05609-29
Call 1-888-358-4705 for availability.
$1,032.00 USD/EACH
Qty:
EW-32610-36 Sample image only.  Check individual details.
Precision Turbine Flowmeter, 0.8 to 6 GPM, 1/2" NPT(M), with display
Product Type Turbine Flowmeters,Description Turbine Flowmeter,Max flow rate GPM 0.8 to 6.0
EW-32610-36
Usually ships in 11 days.
$1,450.00 USD/EACH
Qty:
EW-05610-08 A109GMA200NA2:2in30/300GPM Digital Flowmeter 2inALUMINUM Turbine Housing 2TOTALS 2CALS And Flowrate
A109GMA200NA2:2in30/300GPM Digital Flowmeter 2inALUMINUM Turbine Housing 2TOTALS 2CALS And Flowrate
Product Type Turbine Flowmeters,Description Turbine Flowmeter,Max operating temperature º F 140
EW-05610-08
Usually ships in 7 days.
$983.00 USD/EACH
Qty:
Representative photo only Battery powered turbine flowmeter totalizer 1 Tri Clover 2 to 20 gpm 7 6 to 75 7 LPM 316 SS
Battery-powered turbine flowmeter/totalizer, 1" Tri-Clover, 2 to 20 gpm/7.6 to 75.7 LPM, 316 SS
Product Type Turbine Flowmeters,Description Turbine Flowmeter,Max operating temperature º F 140
EW-05609-11
Usually ships in 14 days.
$836.00 USD/EACH
Qty:
Representative photo only Battery powered turbine flowmeter totalizer 1 2 NPT F 1 to 10 gpm 3 8 to 38 LPM PVDF
Battery-powered turbine flowmeter/totalizer, 1/2" NPT(F), 1 to 10 gpm/3.8 to 38 LPM, PVDF
Product Type Turbine Flowmeters,Description Turbine Flowmeter,Max operating temperature º F 140
EW-05610-11
Call 1-888-358-4705 for availability.
$789.00 USD/EACH
Qty:
257 results (showing 1 - 15 )
Per Page 
View products by: Title | Brand | Category | Price | Popularity | Best Sellers
View product categories by: Popularity | Our Choices | All-Around Favorites | Title | Product Category | Recommended
View popular items by: Popularity | Favorites | Title | Price | Part Number | Category
View Popular Categories by: Popularity | Our Choices | All-Around Favorites | Product Category | Recommended | Title