Mixers and Blenders
pH Meters and Water Quality Products
Pressure & Vacuum Instruments
Process Equipment & Instrumentation
Pumps
Recorders & Data Acquisition
Safety Products
Shop by Industry
Spectroscopy and Spectrophotometers
Temperature, Humidity, Moisture and Weather Products
Tubing, and Hose
Shop by
|
Service & Support
|
Technical Resources
|
My Account
Check order status
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here.
Call
1-800-323-4340
Option
9
ext.
6334
See Details
Home
>
Masterflex Peristaltic Pumps
>
Masterflex Tubing and Pump Accessories
>
Interface Cables, RS-232 / DB9 / USB
> USB to DB9 (M) RS-232 Serial Adapter Cable
USB to DB9 (M) RS-232 Serial Adapter Cable
Use this cable to connect the Masterflex
®
Computer Compatible drives 07550-30, and -50 to a PC with USB port. Will also connect other peripheral devices equipped with RS-232 serial interface including balances, printers, scanners, and PDA’s. Includes 2-ft cable and driver software. Supports data transfer speeds up to 500 Kbps; compatible with Windows
®
98SE, ME, 2000, and XP.
Close
Save My Shopping List
Please login in order to use Shopping Lists
Close
Close
Save My Shopping List
You can
view your shopping list
or
continue shopping
Compare
Product Detail
Sort
Most Popular
Name
Name Descending
Price (Lowest First)
Price (Highest First)
Availability
Price
USB to RS-232 Serial Adapter Cable
Product Type Accessory,Brand Masterflex,Model UR-2000M2
EW-22050-58
In stock.
$39.00 USD/EACH
Qty:
Add to Cart
Save to shopping list
Accessories
Calibrations
More Details
See all
USB to DB9 (M) RS-232 Serial Adapter Cable
1
results (showing
1
-
1
)
Per Page
5
10
15
25
50
Home
>
Masterflex Peristaltic Pumps
>
Masterflex Tubing and Pump Accessories
>
Interface Cables, RS-232 / DB9 / USB
> USB to DB9 (M) RS-232 Serial Adapter Cable
View products by:
Title
|
Brand
|
Category
|
Price
|
Popularity
|
Best Sellers
View product categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Title
|
Product Category
|
Recommended
View popular items by:
Popularity
|
Favorites
|
Title
|
Price
|
Part Number
|
Category
View Popular Categories by:
Popularity