Mixers and Blenders
pH Meters and Water Quality Products
Pressure & Vacuum Instruments
Process Equipment & Instrumentation
Pumps
Recorders & Data Acquisition
Safety Products
Shop by Industry
Spectroscopy and Spectrophotometers
Temperature, Humidity, Moisture and Weather Products
Tubing, and Hose
Shop by
|
Service & Support
|
Technical Resources
|
My Account
Check order status
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here.
Call
1-800-323-4340
Option
9
ext.
6334
See Details
Home
>
Lab Equipment
>
Ultrasonic Cleaners
> Ultrasonic Cavitation Meter
Ultrasonic Cavitation Meter
Instantly measures ultrasonic energy in cleaning tanks
Calculate best workloads
Detect loss of ultrasonic energy
Compare energy level over the life of the tank
Cavitation is the expansion and rapid implosion of microscopic gas or vapor bubbles produced when high-intensity ultrasonic waves are directed into a liquid. Use the meter to determine cavitation intensity in the ultrasonic cleaning tank, to check the effect of work baskets, fixtures, and workloads, and to compare energy levels from tank to tank.
Close
Save My Shopping List
Please login in order to use Shopping Lists
Close
Close
Save My Shopping List
You can
view your shopping list
or
continue shopping
Compare
Product Detail
Sort
Most Popular
Name
Name Descending
Price (Lowest First)
Price (Highest First)
Availability
Price
Ultrasonic Cavitation Meter
Product Type Cavitation Meter,Brand Cavins,Model CM-3-100
EW-08894-60
Call 1-888-358-4705 for availability.
$1,195.00 USD/EACH
Qty:
Add to Cart
Save to shopping list
Accessories
Calibrations
More Details
See all
Ultrasonic Cavitation Meter
1
results (showing
1
-
1
)
Per Page
5
10
15
25
50
Home
>
Lab Equipment
>
Ultrasonic Cleaners
> Ultrasonic Cavitation Meter
View products by:
Title
|
Brand
|
Category
|
Price
|
Popularity
|
Best Sellers
View product categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Title
|
Product Category
|
Recommended
View popular items by:
Popularity
|
Favorites
|
Title
|
Price
|
Part Number
|
Category
View Popular Categories by:
Popularity