Home  >  Lab Equipment > Ultrasonic Cleaners
Ultrasonic Cleaners
Ultrasonic Cleaners
An industrial ultrasonic cleaner uses sounds waves between 20 and 400 kHz to clean objects. A solvent specific to the application is also used. Processing time in an industrial ultrasonic cleaner is normally between two and six minutes. An industrial ultrasonic cleaner usually relies on cavitations caused by the ultrasonic waves to perform the cleaning. Solvents with lower surface tensions will produce greater cleaning action. It is also best to choose a solvent your contaminants are soluble in. In some cases this means an organic solvent. A higher frequency industrial ultrasonic cleaner is generally capable of cleaning with greater detail and a bit more gently. A low frequency one is generally considered a coarse instrument. Typical samples processed by ultrasonic cleaners include: hard metals, ceramics, glass, and plastic. You will find ultrasonic cleaners in a variety of industries such as: automotive, biology, printing, medical, firearms, and semiconductor.
 Product Detail
Sort
 Availability
 Price
EW-08849-00 Cole-Parmer<small><sup>&reg;</sup></small> one-pint compact ultrasonic cleaner, 115 VAC
Cole-Parmer one-pint compact ultrasonic cleaner, 115 VAC
Product Type Cleaning Solutions,Capacity gal. <.5,Brand Cole-Parmer
EW-08849-00
In stock.
$220.00 USD/EACH
Qty:
EW-08895-75 IMAGE IS REPRESENTATIVE OF ITEM.
Cole-Parmer Ultrasonic Cleaner with Digital Timer and Heater, 2-1/2 gallon, 115 VAC
Product Type Ultrasonic Cleaner,Capacity gal. 2.5,Power VAC 115
EW-08895-75
Call 1-888-358-4705 for availability.
$1,370.00 USD/EACH
Qty:
See all
EW-08895-38 Cole-Parmer<small><sup>&reg;</sup></small> Stainless Steel Ultrasonic Cleaner with Heater and Mechanical Timer, 2 1/2 gal, 115 VAC
Cole-Parmer Stainless Steel Ultrasonic Cleaner with Heater and Mechanical Timer, 2 1/2 gal, 115 VAC
Product Type Ultrasonic Cleaner,Capacity gal. 2.5,Power VAC 115
EW-08895-38
Call 1-888-358-4705 for availability.
$1,293.00 USD/EACH
Qty:
EW-08895-91 IMAGE IS REPRESENTATIVE OF ITEM.
Cole-Parmer Ultrasonic Cleaner with Digital Timer and Heater, 5-1/2 gallon, 115 VAC
Product Type Ultrasonic Cleaner,Capacity gal. 5.5,Power VAC 115
EW-08895-91
Call 1-888-358-4705 for availability.
$2,395.00 USD/EACH
Qty:
See all
EW-08895-16 Cole-Parmer<small><sup>&reg;</sup></small> Stainless Steel Ultrasonic Cleaner with Heater and Digital Timer, 1 gal, 115 VAC
Cole-Parmer Stainless Steel Ultrasonic Cleaner with Heater and Digital Timer, 1 gal, 115 VAC
Product Type Ultrasonic Cleaner,Capacity gal. 1,Power VAC 115
EW-08895-16
In stock.
$775.00 USD/EACH
Qty:
Representative photo only Cole Parmer Stainless Steel Ultrasonic Cleaner with Heater and Mechanical Timer 3 1 2 gal 115 VAC
Cole-Parmer Stainless Steel Ultrasonic Cleaner with Heater and Mechanical Timer, 3 1/2 gal, 115 VAC
Product Type Ultrasonic Cleaner,Capacity gal. 3.5,Power VAC 115
EW-08895-50
Call 1-888-358-4705 for availability.
$1,613.00 USD/EACH
Qty:
Representative photo only Cole Parmer Stainless Steel Ultrasonic Cleaner with Heater and Mechanical Timer 3 4 gal 220 VAC
Cole-Parmer Stainless Steel Ultrasonic Cleaner with Heater and Mechanical Timer, 3/4 gal, 220 VAC
Product Type Ultrasonic Cleaner,Capacity gal. 0.75,Power VAC 220
EW-08895-08
Call 1-888-358-4705 for availability.
$613.00 USD/EACH
Qty:
Representative photo only Cole Parmer Stainless Steel Ultrasonic Cleaner with Heater and Mechanical Timer 3 4 gal 115 VAC
Cole-Parmer Stainless Steel Ultrasonic Cleaner with Heater and Mechanical Timer, 3/4 gal, 115 VAC
Product Type Ultrasonic Cleaner,Capacity gal. 0.75,Power VAC 115
EW-08895-02
In stock.
$484.00 USD/EACH
Qty:
Representative photo only Cole Parmer ultrasonic cleaner with timer heater temperature monitor and degas 5 1 2 gallon 230 VAC
EW-08894-26
Call 1-888-358-4705 for availability.
$2,930.00 USD/EACH
Qty:
EW-08894-60 Ultrasonic Cavitation Meter
Ultrasonic Cavitation Meter
Product Type Cavitation Meter,Brand Cavins,Model CM-3-100
EW-08894-60
Call 1-888-358-4705 for availability.
$1,195.00 USD/EACH
Qty:
Representative photo only Cole Parmer Stainless Steel Ultrasonic Cleaner with Heater and Digital Timer 3 4 gal 115 VAC
Cole-Parmer Stainless Steel Ultrasonic Cleaner with Heater and Digital Timer, 3/4 gal, 115 VAC
Product Type Ultrasonic Cleaner,Capacity gal. 0.75,Power VAC 115
EW-08895-04
In stock.
$580.00 USD/EACH
Qty:
Representative photo only Cole Parmer ultrasonic cleaner with timer heater temperature monitor and degas 2 1 2 gallon 230 VAC
EW-08893-26
Call 1-888-358-4705 for availability.
$1,710.00 USD/EACH
Qty:
Representative photo only Cole Parmer Stainless Steel Ultrasonic Cleaner with Heater and Digital Timer 2 1 2 gal 115 VAC
Cole-Parmer Stainless Steel Ultrasonic Cleaner with Heater and Digital Timer, 2 1/2 gal, 115 VAC
Product Type Ultrasonic Cleaner,Capacity gal. 2.5,Power VAC 115
EW-08895-40
Call 1-888-358-4705 for availability.
$1,430.00 USD/EACH
Qty:
Representative photo only Cole Parmer Stainless Steel Ultrasonic Cleaner with Heater and Mechanical Timer 1 gal 115 VAC
Cole-Parmer Stainless Steel Ultrasonic Cleaner with Heater and Mechanical Timer, 1 gal, 115 VAC
Product Type Ultrasonic Cleaner,Capacity gal. 1,Power VAC 115
EW-08895-14
In stock.
$685.00 USD/EACH
Qty:
EW-08895-69 IMAGE IS REPRESENTATIVE OF ITEM.
Cole-Parmer Ultrasonic Cleaner with Mechanical Timer, 2-1/2 gallon, 115 VAC
Product Type Ultrasonic Cleaner,Capacity gal. 2.5,Power VAC 115
EW-08895-69
Call 1-888-358-4705 for availability.
$1,160.00 USD/EACH
Qty:
See all
139 results (showing 1 - 15 )
Per Page 
Home  >  Lab Equipment > Ultrasonic Cleaners
View products by: Title | Brand | Category | Price | Popularity | Best Sellers
View product categories by: Popularity | Our Choices | All-Around Favorites | Title | Product Category | Recommended
View popular items by: Popularity | Favorites | Title | Price | Part Number | Category
View Popular Categories by: Popularity | Our Choices | All-Around Favorites | Product Category | Recommended | Title