This product is now discontinued, View Related Products
Ultrasonic Leak Detector
“Listen” for pressurized gas leaks
  • Detects the ultrasonic noise created by pressure or vacuum leaks of any gas, regardless of type: CFC's, HFC's, nitrogen, CO2, or steam
  • Unaffected by contaminants or windy conditions
  • Identifies failing solenoids, valves, and bearings
  • Includes he
Ultrasonic leak detectors have a decided advantage over conventional leak detectors—they "listen” for leaks and alert you to their presence. There is no gas sensor to become clogged or faulty, and no gas interferences to worry about. This leak detector locates any pressurized gas leak. It is excellent for use with boilers, furnaces, heat exchangers, and chillers. You can also use it to inspect bearings on pumps and machinery, or detect arcing electrical switchgear from a safe distance. Even windy outdoor conditions won’t affect its performance. The high-intensity LED indicator provides good visibility indoors or out. Use the ultrasonic transmitter (sold separately) with the leak detector to amplify leaks in insufficiently pressurized applications. The transmitter is useful in checking gaskets or panes of glass. Ultrasonic waves produce a different sound when a gasket or surface is damaged.
Call For Price
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Gas Measured Range TLV (ppm)
Availability Pricing
Amprobe Ultrasonic Gas Leak Detector EW-86417-00 Mfr # ULD-300
COMPARE
Call For Price
Amprobe Ultrasonic transmitter; for use with ultrasonic leak detector EW-86417-01 Mfr # UT-300