Mixers and Blenders
pH Meters and Water Quality Products
Pressure & Vacuum Instruments
Process Equipment & Instrumentation
Pumps
Recorders & Data Acquisition
Safety Products
Shop by Industry
Spectroscopy and Spectrophotometers
Temperature, Humidity, Moisture and Weather Products
Tubing, and Hose
Shop by
|
Service & Support
|
Technical Resources
|
My Account
Check order status
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here.
Call
1-800-323-4340
Option
9
ext.
6334
See Details
Home
>
Pressure & Vacuum Instruments
>
Vacuum Controllers, Gauges, and Systems
> Vacuum Gauge with Digital Dial and Reading
Vacuum Gauge with Digital Dial and Reading
The two-point factory calibration ensures gauge accuracy for precise readings
Select from three measuring ranges--torr, mbar, and hectopascal
Gauge is ideal for analytical measurements as well as industrial processes. Simply install gauge in-line to start taking measurements--no external sensors to calibrate. The two-point factory calibration ensures gauge accuracy for precise readings.
Gauge features rugged, corrosion-resistant design for demanding applications. The polypropylene housing protects internal components. It's alumina ceramic wetted meterial makes gauge compatible with most corrosives.
Gauge runs on two AA batteries (included). Batteries power gauge for approximately 5 years. Gauge features automatic sleep mode that conserves battery life. Set automatic sleep mode from 0 to 1000 minutes to deactivate display. Gauge includes standard NW16 flange connector and screw on adapter for 1/4" to 3/8" ID tubing.
Close
Save My Shopping List
Please login in order to use Shopping Lists
Close
Close
Save My Shopping List
You can
view your shopping list
or
continue shopping
Compare
Product Detail
Sort
Most Popular
Name
Name Descending
Price (Lowest First)
Price (Highest First)
Availability
Price
Vacuum Gauge with Digital Dial and Reading
Product Type Vacuum gauges/controllers,Display 3.5-digit LCD, 0.40"H with LCD analog dial,Model 682,902.0
EW-07379-20
In stock.
$935.00 USD/EACH
Qty:
Add to Cart
Save to shopping list
Accessories
Calibrations
More Details
See all
Vacuum Gauge with Digital Dial and Reading
1
results (showing
1
-
1
)
Per Page
5
10
15
25
50
Home
>
Pressure & Vacuum Instruments
>
Vacuum Controllers, Gauges, and Systems
> Vacuum Gauge with Digital Dial and Reading
View products by:
Title
|
Brand
|
Category
|
Price
|
Popularity
|
Best Sellers
View product categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Title
|
Product Category
|
Recommended
View popular items by:
Popularity
|
Favorites
|
Title
|
Price
|
Part Number
|
Category
View Popular Categories by:
Popularity