log in or register
Price Range:
Availability:
Custom Ordering Solutions
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here. Call 1-800-323-4340 Option 9 ext. 6334
Home  >  Fittings > Gaskets > Sanitary Gaskets > Sanitary Screen Gaskets > Viton screen gaskets
Viton screen gaskets
Viton screen gaskets
  • Protect your pump, valves and other components in fluid line
  • Pre-filter before critical membrane filters
  • Generate turbulence for enhanced mass or heat transfer
  • Purify in chromatography applications
Screen gaskets are designed to be interchangeable with standard sanitary clamp gaskets. Gaskets are available in platinum-cured silicone, Viton®, EPDM, PTFE, and Tuf-Steel in mesh sizes of 10, 20, 40, 60 and 100. Screen mesh is constructed of 316L SS. Gaskets meet USP Class VI, FDA CFR Title 21 Section 177.2600, USDA, 3-A and cGMP criteria. All gaskets are animal-derived ingredient free. Each gasket sold individually.
Viton® gaskets have excellent chemical resistance to many acids and alkalies. Minimize your downtime—Viton gaskets are soft and easy to install. Resistance to hardening and embrittlement prevents leakage even after prolonged installation. Viton gaskets are perfect for high temperature applications from –25 to 400degF (–32 to 204deg
 Product Detail
Sort
 Availability
 Price
Representative photo only Viton Gasket 100 MESH Size 3 4 Tri Clamp
Viton Gasket, 100 MESH, Size 3/4" Tri-Clamp
Product Type Gaskets fittings,Clamp size 0.75,Material Viton
EW-30536-82
Usually ships in 14 days.
$38.00 USD/EACH
Qty:
Representative photo only Viton Gasket 10 MESH Size 1 1 2 Tri Clamp
Viton Gasket, 10 MESH, Size 1-1/2" Tri-Clamp
Product Type Gaskets fittings,Clamp size 1.5,Material Viton
EW-30536-06
In stock.
$36.50 USD/EACH
Qty:
Representative photo only Viton Gasket 100 MESH Size 1 1 2 Tri Clamp
Viton Gasket, 100 MESH, Size 1-1/2" Tri-Clamp
Product Type Gaskets fittings,Clamp size 1.5,Material Viton
EW-30536-86
In stock.
$38.00 USD/EACH
Qty:
Representative photo only Viton Gasket 100 MESH Size 2 Tri Clamp
Viton Gasket, 100 MESH, Size 2" Tri-Clamp
Product Type Gaskets fittings,Clamp size 2,Material Viton
EW-30536-88
In stock.
$40.50 USD/EACH
Qty:
Representative photo only Viton Gasket 40 MESH Size 1 1 2 Tri Clamp
Viton Gasket, 40 MESH, Size 1-1/2" Tri-Clamp
Product Type Gaskets fittings,Clamp size 1.5,Material Viton
EW-30536-46
Usually ships in 13 days.
$38.00 USD/EACH
Qty:
Representative photo only Viton Gasket 100 MESH Size 1 2 Tri Clamp
Viton Gasket, 100 MESH, Size 1/2" Tri-Clamp
Product Type Gaskets fittings,Clamp size 0.5,Material Viton
EW-30536-80
Usually ships in 10 days.
$38.00 USD/EACH
Qty:
Representative photo only Viton Gasket 40 MESH Size 3 4 Tri Clamp
Viton Gasket, 40 MESH, Size 3/4" Tri-Clamp
Product Type Gaskets fittings,Clamp size 0.75,Material Viton
EW-30536-42
Usually ships in 5 days.
$38.00 USD/EACH
Qty:
Representative photo only Viton Gasket 40 MESH Size 2 Tri Clamp
Viton Gasket, 40 MESH, Size 2" Tri-Clamp
Product Type Gaskets fittings,Clamp size 2,Material Viton
EW-30536-48
Usually ships in 9 days.
$40.50 USD/EACH
Qty:
Representative photo only Viton Gasket 20 MESH Size 2 Tri Clamp
Viton Gasket, 20 MESH, Size 2" Tri-Clamp
Product Type Gaskets fittings,Clamp size 2,Material Viton
EW-30536-28
Usually ships in 13 days.
$40.00 USD/EACH
Qty:
Representative photo only Viton Gasket 60 MESH Size 1 1 2 Tri Clamp
Viton Gasket, 60 MESH, Size 1-1/2" Tri-Clamp
Product Type Gaskets fittings,Clamp size 1.5,Material Viton
EW-30536-66
Usually ships in 4 days.
$38.00 USD/EACH
Qty:
Representative photo only Viton Gasket 60 MESH Size 2 Tri Clamp
Viton Gasket, 60 MESH, Size 2" Tri-Clamp
Product Type Gaskets fittings,Clamp size 2,Material Viton
EW-30536-68
Usually ships in 7 days.
$40.50 USD/EACH
Qty:
Representative photo only Viton Gasket 10 MESH Size 2 Tri Clamp
Viton Gasket, 10 MESH, Size 2" Tri-Clamp
Product Type Gaskets fittings,Clamp size 2,Material Viton
EW-30536-08
In stock.
$40.00 USD/EACH
Qty:
Representative photo only Viton Gasket 60 MESH Size 1 Tri Clamp
Viton Gasket, 60 MESH, Size 1" Tri-Clamp
Product Type Gaskets fittings,Clamp size 1,Material Viton
EW-30536-64
Usually ships in 6 days.
$38.00 USD/EACH
Qty:
Representative photo only Viton Gasket 20 MESH Size 1 Tri Clamp
Viton Gasket, 20 MESH, Size 1" Tri-Clamp
Product Type Gaskets fittings,Clamp size 1,Material Viton
EW-30536-24
Usually ships in 8 days.
$38.00 USD/EACH
Qty:
Representative photo only Viton Gasket 10 MESH Size 1 Tri Clamp
Viton Gasket, 10 MESH, Size 1" Tri-Clamp
Product Type Gaskets fittings,Clamp size 1,Material Viton
EW-30536-04
Usually ships in 7 days.
$36.50 USD/EACH
Qty:
23 results (showing 1 - 15 )
Per Page 
Home  >  Fittings > Gaskets > Sanitary Gaskets > Sanitary Screen Gaskets > Viton screen gaskets
View products by: Title | Brand | Category | Price | Popularity | Best Sellers
View product categories by: Popularity | Our Choices | All-Around Favorites | Title | Product Category | Recommended
View popular items by: Popularity | Favorites | Title | Price | Part Number | Category
View Popular Categories by: Popularity | Our Choices | All-Around Favorites | Product Category | Recommended | Title