United States
[Change]
Shopping Cart
Subtotal
=
$0.00
Items
=
0
View Cart
United States
1-800-323-4340
or
Live Chat
Welcome,
log in
or
register
Search
Shop All Products
Balances & Scales
Baths and Circulators
Calibration
Chemicals, Reagents, & Lubricants
Clean Room
Cooling Equipment
Fittings
Flow, Level & Valves
Fluid & Liquid Handling
Furniture & Fume Hoods
Heating Equipment
Industrial Equipment
Lab Equipment
Laboratory Cleaning Supplies
Laboratory Supplies & Consumables
Life Science
Masterflex Peristaltic Pumps
Masterflex Pump Tubing
Material Handling and Storage
Material Testing & Physical Testing
Mixers and Blenders
pH Meters and Water Quality Products
Pressure & Vacuum Instruments
Process Equipment & Instrumentation
Pumps
Recorders & Data Acquisition
Safety Products
Shop by Industry
Spectroscopy and Spectrophotometers
Temperature, Humidity, Moisture and Weather Products
Tubing, and Hose
Shop by
|
Service & Support
|
Technical Resources
|
My Account
Check order status
rpm:
1750
(1)
1450
(1)
Leasing Available
Financing is avaliable for many of our products.
See Details
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here.
Call
1-800-323-4340
Option
9
ext.
6334
See Details
Home
>
Lab Equipment
>
Sieves
>
Sieve Shakers
>
Ro-tap, Ro-tap II, and Ro-tap E Sieve Shakers
> W.S. Tyler Ro-Tap E Sieve Shakers
W.S. Tyler Ro-Tap E Sieve Shakers
Reproduce the rotating and tapping motion of hand-sieving but with more uniform motion for highly reproducible results
Ro-Tap
®
E Sieve Shakers for 8'' dia sieves hold up to six full-height or 13 half-height sieves with top cover and receiving pan
Shakers are actuated by a built-in, 99-minute digital timer with 0.1 second accuracy. They are powered by a 1/4-hp motor and include a 5-ft cord (110 VAC models also include U.S. three-prong plug).
Ro-Tap
®
E Sieve Shakers for 8'' sieves are stand-alone benchtop units that are virtually maintenance-free, offer quiet operation and can be operated directly on a laboratory countertop. Options include automatic controlled amplitude, and fine and coarse analysis settings.
Close
Save My Shopping List
Please login in order to use Shopping Lists
Close
Close
Save My Shopping List
You can
view your shopping list
or
continue shopping
Compare
Product Detail
Sort
Most Popular
Name
Name Descending
Price (Lowest First)
Price (Highest First)
Availability
Price
W.S. Tyler Ro-Tap E Sieve Shaker, for 8" diameter sieves, 110 VAC, 60 Hz
Product Type Sieve Shakers,Sieves size accepted " OD 8",rpm 1750
EW-59996-68
Usually ships in 14 days.
$4,410.00 USD/EACH
Qty:
Add to Cart
Save to shopping list
Accessories
Calibrations
More Details
See all
W.S. Tyler Ro-Tap E Sieve Shakers
W.S. Tyler Ro-Tap E Sieve Shaker, for 8" diameter sieves, 220 VAC, 50 Hz
Product Type Sieve Shakers,Sieves size accepted " OD 8,rpm 1450
EW-59996-70
Usually ships in 14 days.
$5,011.00 USD/EACH
Qty:
Add to Cart
Save to shopping list
Accessories
Calibrations
More Details
See all
W.S. Tyler Ro-Tap E Sieve Shakers
2
results (showing
1
-
2
)
Per Page
5
10
15
25
50
Home
>
Lab Equipment
>
Sieves
>
Sieve Shakers
>
Ro-tap, Ro-tap II, and Ro-tap E Sieve Shakers
> W.S. Tyler Ro-Tap E Sieve Shakers
View products by:
Title
|
Brand
|
Category
|
Price
|
Popularity
|
Best Sellers
View product categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Title
|
Product Category
|
Recommended
View popular items by:
Popularity
|
Favorites
|
Title
|
Price
|
Part Number
|
Category
View Popular Categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Product Category
|
Recommended
|
Title