Subtotal = $0.00 
Items = 0 
View Cart
United States
1-800-323-4340 or 
Welcome,
log in or register
Price Range:
Quantity (mL):
500 (1)
120 (1)
Custom Ordering Solutions
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here. Call 1-800-323-4340 Option 9 ext. 6334
 
Home  >  Chemicals, Reagents, & Lubricants > Water > Water Hardness Indicator

Water Hardness Indicator

Water Hardness Indicator
Choose from our comprehensive line of Water Hardness Indicator and other Water products.
 Product Detail
Sort
 Availability
 Price
Representative photo only Water Hardness Indicator 1 w v Eriochrome Black T in 20 v v Triethanolamine 80 v v Isopropyl Alcohol 500 mL
EW-86977-17
Usually ships in 15 days.
$33.00 USD/EACH

Qty:

Representative photo only Water Hardness Indicator 1 w v Eriochrome Black T in 20 v v Triethanolamine 80 v v Isopropyl Alcohol 120 mL
EW-86992-77
Usually ships in 14 days.
$18.00 USD/EACH

Qty:

2 results (showing 1 - 2 )
Per Page 
Home  >  Chemicals, Reagents, & Lubricants > Water > Water Hardness Indicator

View products by: Title | Brand | Category | Price | Popularity | Best Sellers
View product categories by: Popularity | Our Choices | All-Around Favorites | Title | Product Category | Recommended
View popular items by: Popularity | Favorites | Title | Price | Part Number | Category
View Popular Categories by: Popularity | Our Choices | All-Around Favorites | Product Category | Recommended | Title