Mixers and Blenders
pH Meters and Water Quality Products
Pressure & Vacuum Instruments
Process Equipment & Instrumentation
Pumps
Recorders & Data Acquisition
Safety Products
Shop by Industry
Spectroscopy and Spectrophotometers
Temperature, Humidity, Moisture and Weather Products
Tubing, and Hose
Shop by
|
Service & Support
|
Technical Resources
|
My Account
Check order status
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here.
Call
1-800-323-4340
Option
9
ext.
6334
See Details
Home
>
Industrial Equipment
>
Tools
>
Soldering Tools
> Weller Professional Butane Soldering Pen Kit
Weller Professional Butane Soldering Pen Kit
Soldering pen equivalent to a 25 to 75 watt soldering iron
ESD safe to protect sensitive components
Pen has an automatic cut-off switch when the cap is replaced
This lightweight soldering pen features professional grade quality in a compact design. Operate as a soldering iron or a heat tool. Gas catalyst system produces predictable and repeatable temperatures to ensure professional results. Convenient push-button switch offers quick and convenient ignition. Torch is powered by butane; order separately below.
Close
Save My Shopping List
Please login in order to use Shopping Lists
Close
Close
Save My Shopping List
You can
view your shopping list
or
continue shopping
Compare
Product Detail
Sort
Most Popular
Name
Name Descending
Price (Lowest First)
Price (Highest First)
Availability
Price
Weller professional butane soldering pen kit
Product Type Self Igniting,Description Weller Professional Butane Solder Kit,Model 037103169372
EW-97105-61
Usually ships in 10 days.
$95.00 USD/EACH
Qty:
Add to Cart
Save to shopping list
Accessories
Calibrations
More Details
See all
Weller Professional Butane Soldering Pen Kit
1
results (showing
1
-
1
)
Per Page
5
10
15
25
50
Home
>
Industrial Equipment
>
Tools
>
Soldering Tools
> Weller Professional Butane Soldering Pen Kit
View products by:
Title
|
Brand
|
Category
|
Price
|
Popularity
|
Best Sellers
View product categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Title
|
Product Category
|
Recommended
View popular items by:
Popularity
|
Favorites
|
Title
|
Price
|
Part Number
|
Category
View Popular Categories by:
Popularity