Subtotal = $0.00 
Items = 0 
View Cart
United States
1-800-323-4340 or 
Welcome,
log in or register
Category:
Brand:
Whatman (14)
Product Type:
Availability:
In Stock (104)
CE Compliance:
Yes (29)
No. of wells:
10 (25)
14 (20)
12 (12)
20 (11)
27 (6)
16 (5)
15 (3)
8/10 (3)
12/16 (3)
10/14 (3)
8/16 (3)
13 (3)
26 (3)
28 (3)
51 (3)
102 (3)
30 (3)
4 (2)
Buffer solution required (mL):
1600 (16)
250 (3)
300 (3)
2300 (2)
700 (2)
400 (1)
1000 (1)
4500 (1)
1400 (1)
450 (1)
3000 (1)
150 (1)
750 (1)
1500 (1)
725 (1)
500 (1)
825 (1)
900 (1)
3800 (1)
1100 (1)
Gel thickness (mm):
1.5 (15)
0.4 (1)
Gel size (cm L):
8 (10)
25 (4)
10 (3)
40 (1)
45 (1)
8.5 (1)
16 (1)
20 (1)
26 (1)
Gel size (cm W):
8.5 (9)
20 (3)
14 (3)
13 (2)
7.5 (2)
7 (1)
23 (1)
5.5 (1)
26 (1)
Glass size (cm H):
10 (9)
8 (9)
14.5 (7)
16 (7)
19 (7)
22 (7)
28 (7)
Glass size (cm W):
16.5 (9)
20 (6)
16 (3)
Qty/pk:
100 (6)
1 (3)
Glass size (cm L):
20 (5)
14 (3)
45 (1)
14.5 (1)
16 (1)
19 (1)
22 (1)
28 (1)
Ash:
0.06% (4)
0.01% (2)
Pore size (micron):
0.45 (2)
0.2 (1)

Learn more about
Western Blotting Products

2,600 pages
of answers

FREE
2013/14 Cole-Parmer
Sourcebook
Choose from over 250 pages of products for life science. Choose from a number of well known brands including Corning, Thermo Scientific, UVP, EMD Chemicals, Nalge NUNC, and Eppendorf.
Request your copy today.

Leasing Available
Financing is avaliable for many of our products.

See Details
Custom Ordering Solutions
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here. Call 1-800-323-4340 Option 9 ext. 6334

More Unique Lab & Industrial Supplies

 

Western Blotting

Western Blotting

Cole-Parmer offers a complete line of products for Western Blotting including horizontal and vertical gel electrophoresis systems, reagents, electroblotters, imaging systems, and spectrophotometers. Along with our wide range of products, we also offer expert assistance in finding the right product. For more information or help choosing the right product for your need, contact our Application Specialists.

Western blot is a technique utilized for the detection of proteins within a tissue sample. Testing for HIV and mad cow disease utilize Western blotting as well. The overall Western blot process is:

Still have questions?
Contact Our Experts
for FREE product
application selection
and support.
or call 1-800-323-4340
 Product Detail
Sort
 Availability
 Price
Representative photo only Cole Parmer Blue Sensitive Autoradiography Film 8 x 10 100 pk

Cole-Parmer Blue-Sensitive Autoradiography Film, 8 x 10", 100/pk

Product Type Electrophoresis Accessories,Brand Cole-Parmer
EW-21700-03
In stock.
$100.00 USD/PACKAGE

Qty:

Representative photo only Instant Blue Stain 1 liter

Instant Blue Stain, 1 liter

Product Type Electrophoresis Accessories,Product Type Electrophoresis Accessories,No. of wells 20
EW-28570-59
In stock.
$209.00 USD/EACH

Qty:

EW-97700-20 PhotoDoc-It 50 Imaging System is shown here for illustrative purposes only. Please refer to product description.

PhotoDoc-It Imaging System with 302/365nm 20cm x 20cm Filter Transilluminator, 230VAC

Product Type Electrophoresis Imaging Systems,Brand PhotoDoc-It
EW-97700-20
Usually ships in 10 days.
$3,944.00 USD/EACH

Qty:

EW-06645-80 Nitrocellulose membranes 0.45um 300cm/roll

Nitrocellulose membranes 0.45um 300cm/roll

Product Type Nitrocellulose Membranes,Filter size 0.45 um,Pore size micron 0.45
EW-06645-80
In stock.
$230.00 USD/ROLL

Qty:

Representative photo only Cole Parmer 8 Watt Variable Intensity UV Transilluminator 302nm Wavelength with 21cm x 26cm Filter 230VAC
EW-97623-18
Usually ships in 10 days.
$1,764.00 USD/EACH

Qty:

EW-97700-10 PhotoDoc-It 50 Imaging System is shown here for illustrative purposes only. Please refer to product description.

PhotoDoc-It 50 Imaging System, 115VAC

Product Type Electrophoresis Imaging Systems,Brand PhotoDoc-It
EW-97700-10
Usually ships in 14 days.
$2,248.00 USD/EACH

Qty:

EW-28560-00 Midi-horizontal gel units, 14 x 10 cm gel bed

Midi-horizontal gel units, 14 x 10 cm gel bed

Product Type Electrophoresis Horizontal Systems,Buffer solution required mL 700,No. of wells 10, 13, 16, 20, 27
EW-28560-00
Call 1-888-358-4705 for availability.
$385.00 USD/EACH

Qty:

Representative photo only X RAY FILM CASSETTE STAINLESS STEEL 8X10
EW-21700-43
Usually ships in 3 days.
$305.00 USD/EACH

Qty:

Representative photo only Thermo Scientific Electrophoresis Power Supply 1000v 230v UK plug

Thermo Scientific Electrophoresis Power Supply 1000v 230v UK plug

Dimensions " W 5-½"W,Dimensions " H 5-¼"H,Dimensions " D 10-¼"L
EW-28406-35
Call 1-888-358-4705 for availability.
$1,532.00 USD/EACH

Qty:

EW-28562-00 Cole-Parmer<small><sup>&reg;</sup></small> High-Throughput Horizontal Gel System

Cole-Parmer High-Throughput Horizontal Gel System

Product Type Electrophoresis Horizontal Systems,Buffer solution required mL 3000,No. of wells 26, 27, 28, 51, 102
EW-28562-00
Call 1-888-358-4705 for availability.
$522.00 USD/EACH

Qty:

Representative photo only Doc It LS Imaging Software order in addition to BioDoc It Imaging Systems

Doc-It LS Imaging Software- order in addition to BioDoc-It Imaging Systems

Product Type Electrophoresis Imaging Systems Accessories,Brand UVP
EW-97701-62
Usually ships in 14 days.
$1,669.00 USD/EACH

Qty:

Representative photo only Thermo Scientific Electrophoresis Power Supply 300v 230v European plug

Thermo Scientific Electrophoresis Power Supply 300v 230v European plug

Dimensions " W 5-½"W,Dimensions " H 5-¼"H,Dimensions " D 10-¼"L
EW-28406-25
Usually ships in 8 days.
$619.00 USD/EACH

Qty:

EW-17314-11 Enduro Gel XL Electrophoresis System 120V

Enduro Gel XL Electrophoresis System 120V

Product Type Electrophoresis Horizontal System,Buffer solution required mL 300,Dimensions " D 9.625
EW-17314-11
Usually ships in 5 days.
$629.50 USD/EACH

Qty:

Representative photo only X ray Film Blue 5x7 100 pk
EW-21700-01
In stock.
$70.00 USD/PACKAGE

Qty:

Representative photo only Enduro Gel XL Electrophoresis System with UK plug 230V

Enduro Gel XL Electrophoresis System with UK plug 230V

Product Type Electrophoresis Horizontal System,Buffer solution required mL 300,Dimensions " D 9.625
EW-17314-13
Usually ships in 5 days.
$629.50 USD/EACH

Qty:

406 results (showing 1 - 15 )
Per Page 

View products by: Title | Brand | Category | Price | Popularity | Best Sellers
View product categories by: Popularity | Our Choices | All-Around Favorites | Title | Product Category | Recommended
View popular items by: Popularity | Favorites | Title | Price | Part Number | Category
View Popular Categories by: Popularity | Our Choices | All-Around Favorites | Product Category | Recommended | Title