Item Total = $0.00 
Items = 0 
View Cart
United States
1-800-323-4340 or 
Welcome,
log in or register
Custom Ordering Solutions
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here. Call 1-800-323-4340 Option 8 ext. 138-8456
pH Buffers
pH Buffers
Always choose a pH 7 buffer plus at least one other pH value close to your expected measurement range. Using solutions of known pH value allow you to adjust the system to read accurate measurements. Calibration should be performed before each reading or each set of readings.
pH buffers allow you to adjust your testing system to measure your samples' unknown pH more precisely. For best accuracy:
  • Peform standardizations with fresh buffer solutions. Never pour used buffer back into the bottle; always discard after using
  • Use buffers that frame the pH range of the samples you are testing (i.e. directly above and below the expected value of your sample
  • Match the temperature of the buffer to the temperature of the samples. For example, if your samples are at 50 degrees Celcius, warm your buffers to 50 degrees Celcius using a beaker in a water bath.
  • Calibrate your system before each use or set of uses
  • Don't miss this end of year sale!
     Product Detail
    Sort
     Availability
     Price
    Oakton buffer pack, 500 mL each of 4.01, 7.00, and 10.00 buffer
    • Product Type: pH Buffers
    • Volume: 500 mL
    • Qty/pk: 3
    EW-05942-10
    In stock.
    $30.25 USD/PACKAGE of 3
    $19.99 USD/PACKAGE of 3
    Qty:
    Oakton pH Calibration Buffer, 7.00, 500 mL
    • Description: pH 7.00
    • Product Type: pH Buffers
    • Volume: 500 mL
    EW-00654-04
    In stock.
    $13.00 USD/PINT
    Qty:
    Oakton pH Calibration Buffer, 4.01, 500 mL
    • Description: pH 4.01
    • Product Type: pH Buffers
    • Volume: 500 mL
    EW-00654-00
    In stock.
    $13.00 USD/PINT
    Qty:
    Oakton pH Calibration Buffer, 7.00, 1 Gal
    • Description: pH 7.00
    • Product Type: pH Buffers
    • Volume: 4000 mL
    EW-05942-44
    In stock.
    $44.25 USD/EACH
    Qty:
    Oakton pH Calibration Buffer, 4.01, 1 Gal
    • Description: pH 4.01
    • Product Type: pH Buffers
    • Volume: 4000 mL
    EW-05942-24
    In stock.
    $44.25 USD/EACH
    Qty:
    Oakton pH Calibration Buffer, 10.00, 500 mL
    • Description: pH 10.00
    • Product Type: pH Buffers
    • Volume: 500 mL
    EW-00654-08
    In stock.
    $13.00 USD/PINT
    Qty:
    Oakton pH Calibration Buffer, 7.00, 1.0 L
    • Description: pH 7.00
    • Product Type: pH Buffers
    • Volume: 1000 mL
    EW-05942-42
    In stock.
    $22.25 USD/EACH
    Qty:
    Oakton pH Calibration Buffer, 10.00, 1 Gal
    • Description: pH 10.00
    • Product Type: pH Buffers
    • Volume: 4000 mL
    EW-05942-64
    In stock.
    $44.25 USD/EACH
    Qty:
    Oakton pH Calibration Buffer, 4.01, 1.0 L
    • Description: pH 4.01
    • Product Type: pH Buffers
    • Volume: 1000 mL
    EW-05942-22
    In stock.
    $22.25 USD/EACH
    Qty:
    Oakton pH buffer pouches; pH 7.00, 20/pk
    • Description: pH 7.00
    • Product Type: pH Buffers
    • Qty/pk: 20
    EW-35653-02
    In stock.
    $33.25 USD/PACKAGE of 20
    Qty:
    Oakton pH Calibration Buffer, 10.00, 1.0 L
    • Description: pH 10.00
    • Product Type: pH Buffers
    • Volume: 1000 mL
    EW-05942-62
    In stock.
    $22.25 USD/EACH
    Qty:
    Oakton pH Calibration Buffer Pouches, assortment
    • Description: Assortment of pH 4.01, 7.00, and 10.00 buffers and rinse solution
    • Product Type: pH Buffers
    • Qty/pk: 20
    EW-35653-04
    In stock.
    $33.25 USD/PACKAGE of 20
    Qty:
    Oakton pH buffer pouches; pH 4.01, 20/pk
    • Description: pH 4.01
    • Product Type: pH Buffers
    • Qty/pk: 20
    EW-35653-01
    In stock.
    $33.25 USD/PACKAGE of 20
    Qty:
    Oakton pH Solution Pack; pH 4.01/7.00/10.00, Storage/Cleaning, 500 mL each
    • Product Type: pH Buffers
    • Volume: 500 mL
    • Qty/pk: 5
    EW-05942-15
    In stock.
    $57.50 USD/PACKAGE
    Qty:
    Oakton pH buffer pouches; pH 10.00, 20/pk
    • Description: pH 10.00
    • Product Type: pH Buffers
    • Qty/pk: 20
    EW-35653-03
    In stock.
    $33.25 USD/PACKAGE of 20
    Qty:
    105 results (showing 1 - 15 )
    Per Page 
    #PRODUCTTITLE#
    #PRODUCTHEADER#
    #IAMGESTICKER#
    #CROSSOUTPRICE#
    #PRICE#
    #PRICEWITHVAT#
    #AVAILABILITY#
    Qty
    Quantity Discounts
    Qty: #QTY10COUNT# #QTY15COUNT# #QTY20COUNT#
    Price: #QTY10PRICE# #QTY15PRICE# #QTY20PRICE#
    #PRODUCTIMAGE#
    #POWERREVIEW#
    #MARKETINGTEXT#
    #ICONS#
    #SPECS#