Item Total = $0.00 
Items = 0 
View Cart
United States
1-800-323-4340 or 
Welcome,
log in or register
Category:
Brand:
GLI (7)
Product Type:
Availability:
In Stock (55)
Shop Items By:
Material:
PVDF (17)
PEEK (2)
Custom Ordering Solutions
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here. Call 1-800-323-4340 Option 8 ext. 138-8456
 
Home  >  pH/Ion Industrial Electrodes

pH/Ion Industrial Electrodes

pH/Ion Industrial Electrodes
Choose from our comprehensive line of Industrial pH and ORP Electrodes, Assemblies, and Accessories and other pH Electrodes products.
 Product Detail
Sort
 Availability
 Price

Cole-Parmer self-cleaning pH electrode, CPVC housing, 10-ft cable, BNC

  • Product Type: pH Electrode
  • Junction: Double
  • pH range: 0 to 12
EW-27003-00
In stock.
$137.00 USD/EACH

Qty:

Cole-Parmer Submersible 3' L pH Probe, DJ/ABS/None; None/BNC

  • Product Type: pH Electrode
  • Applications: Dirty samples, sulfates, contaminants
  • Junction: Double
EW-27001-80
In stock.
$173.00 USD/EACH

Qty:

Cole-Parmer self-cleaning pH electrode, CPVC housing, 100Ohm Pt RTD, tinned end

  • Product Type: pH Electrode
  • Junction: Double
  • pH range: 0 to 12
EW-27003-12
In stock.
$175.00 USD/EACH

Qty:

pH Electrode, flat surface, Pt-1000 ATC, 3/4" NPT threads

  • Product Type: pH Electrode
  • pH range: 0 to 14
  • Temp range: -10 to 85C
EW-56561-00
In stock.
$340.00 USD/EACH

Qty:

Cole-Parmer Self-Cleaning Flat Surface ORP Probe, DJ/CPVC/Pt/10'/BNC

  • Product Type: ORP Electrode
  • Housing: CPVC
  • Qty/ea: 1
EW-27003-40
In stock.
$184.00 USD/EACH

Qty:

+GF+ Signet DryLoc Preamp with 3/4" NPT threads, 15-ft cable

  • Product Type: Electrode cable
  • Thread size: 3/4"NPT
  • Cable length: 15 ft
EW-56560-03
In stock.
$310.00 USD/EACH

Qty:

pH Electrode, flat surface, 3KOhm Balco thermistor ATC, 3/4" NPT threads

  • Product Type: pH Electrode
  • pH range: 0 to 14
  • Temp range: -10 to 85C
EW-56561-02
In stock.
$340.00 USD/EACH

Qty:

Cole-Parmer Solution Grounded pH Probe, DJ/PPS/100Ohm RTD; Plug/BNC

  • Product Type: pH Electrode
  • Applications: Minimal salt applications
  • Junction: Double
EW-27018-02
In stock.
$281.00 USD/EACH

Qty:

Cole-Parmer General Purpose pH electrode, double-junction, 10-ft cable

  • Product Type: pH Electrode
  • Junction: Double
  • pH range: up to 13
EW-05994-22
Usually ships in 10 days or sooner.
$140.00 USD/EACH

Qty:

Cole-Parmer Submersible 3' L pH Probe, SJ/ABS/None; None/BNC

  • Product Type: pH Electrode
  • Applications: Clean water, general purpose
  • Junction: Single
EW-05993-80
In stock.
$153.00 USD/EACH

Qty:

Cole-Parmer self-cleaning pH electrode, CPVC housing, 100Ohm Pt RTD, mini audio

  • Product Type: pH Electrode
  • Junction: Double
  • pH range: 0 to 12
EW-27003-02
In stock.
$172.00 USD/EACH

Qty:

Cole-Parmer self-cleaning pH electrode, Ryton housing, 10-ft cable, BNC

  • Product Type: pH Electrode
  • Junction: Double
  • pH range: 0 to 12
EW-27003-20
In stock.
$175.00 USD/EACH

Qty:

Cole-Parmer Solution Grounded High-Na pH Probe, DJ/PPS/100Ohm RTD; Tinned/BNC

  • Product Type: pH Electrode
  • Applications: High-salt applications
  • Junction: Double
EW-27018-23
In stock.
$288.00 USD/EACH

Qty:

ORP Electrode, flat surface, 3/4" NPT threads

  • Product Type: ORP Electrode
  • Range: -2000 to 2000 mV
  • Temp range: -10 to 85C
EW-56561-16
Available for order
or call 1-888-358-4705 to order
$394.00 USD/EACH

Qty:

Cole-Parmer General Purpose pH Electrode, 10-ft Cable

  • Product Type: pH Electrode
  • Junction: Single
  • Glass type: Blue
EW-05994-20
In stock.
$115.00 USD/EACH

Qty:

283 results (showing 1 - 15 )
Per Page 
#PRODUCTTITLE#

#PRODUCTHEADER#

#IAMGESTICKER#
#CROSSOUTPRICE#
#PRICE#
#PRICEWITHVAT#
#AVAILABILITY#
Qty
Quantity Discounts
Qty: #QTY10COUNT# #QTY15COUNT# #QTY20COUNT#
Price: #QTY10PRICE# #QTY15PRICE# #QTY20PRICE#
#PRODUCTIMAGE#
#POWERREVIEW#
#MARKETINGTEXT#
#ICONS#
#SPECS#
Home  >  pH/Ion Industrial Electrodes