Home  >  pH/Ion Meter Accessories
pH/Ion Meter Accessories
pH/Ion Meter Accessories
BNC extension cables make it easier to measure pH when a sample is more than few feet away from the pH meter and a handheld pH meter just isn’t practical. Though BNC connectors are the most popular pH meter and electrode connection, there are still meters in use with other connectors like US standard, DIN, Metrohm, Radiometer-type connectors. For those you need an adapter if you plan on using BNC extension cables to connect any pH electrode with a BNC connector to these meters. For distances over 25 feet, as is common in industrial applications with a pH meter, a pH controller or a pH transmitter, BNC extension cables are available with a preamplifier to minimize signal loss. These pH accessories are available up to 100 feet in length. Cole-Parmer also offers preamplified pH electrode cables with automatic temperature compensation (ATC) connections and solution grounds for greater accuracy and more stability in applications where temperatures change or electrical interference for other equipment is common. If you already have an amplifier or just want added signal protection, we recommend heavy-duty pH electrode extension cables with shielded, heavy-duty insulation, 5 mm OD cables, and rubber-hooded BNC connector. For help with BNC extension cables contact a Technical Expert today.
 Product Detail
Sort
 Availability
 Price
Representative photo only  All in one combination pH Temp electrode 3 ft cable epoxy body single junction gel filled sub miniature audio ATC BNC 12 mm dia
EW-35811-71
In stock.
$85.00 USD/EACH
Qty:
Representative photo only  All in one combination pH Temp electrode 3 ft cable epoxy body double junction gel filled sub miniature audio ATC BNC 12 mm dia
EW-35811-72
In stock.
$125.00 USD/EACH
Qty:
Representative photo only Replacement double junction sensor for Oakton waterproof pH Testrs 3 3 10 20 30
EW-35624-38
In stock.
$70.00 USD/EACH
Qty:
EW-35608-50 Ultem Body, SS Electrodes Conductivity Cell for OAKTON<small><sup>&reg;</sup></small> Conductivity and TDS Meters, K=1.0, 2.5' Cable
EW-35608-50
In stock.
$131.00 USD/EACH
Qty:
Representative photo only Replacement pH Con Temp electrode w 10 ft cable for Oakton PC10 meter
EW-35630-50
In stock.
$220.00 USD/EACH
Qty:
EW-59002-75 Replacement rugged "All-in-One" electrode for waterproof meters only, double junction, polymer-filled. Epoxy body, watertight BNC connector.
EW-59002-75
In stock.
$118.00 USD/EACH
Qty:
EW-59500-81 Accumet<small><sup>&reg;</sup></small> Benchtop Meter Replacement pH/ATC Probe
Accumet Benchtop Meter Replacement pH/ATC Probe
Product Type Accessory,Model CP-620-530,Brand Accumet
EW-59500-81
In stock.
$174.00 USD/EACH
Qty:
Representative photo only Replacement Waterproof Combination pH Electrode
Replacement Waterproof Combination pH Electrode
Product Type pH Electrode,Model 103-635
EW-58900-55
In stock.
$235.00 USD/EACH
Qty:
Representative photo only pH Electrode Double junction Sealed Epoxy Body 12 mm Diameter 3 ft Cable BNC Connection
EW-35641-51
In stock.
$84.00 USD/EACH
Qty:
EW-35425-50 Replacement pH/Conductivity/TDS Sensor Module
Replacement pH/Conductivity/TDS Sensor Module
Product Type Conductivity Sensor
EW-35425-50
In stock.
$77.00 USD/EACH
Qty:
Representative photo only Oakton Flexible Benchtop Electrode Holder
EW-35617-50
In stock.
$106.00 USD/EACH
Qty:
Representative photo only Replacement Conductivity Temp probe K 1 0 for Oakton 6 and 6 series meters
EW-35606-55
In stock.
$155.00 USD/EACH
Qty:
Representative photo only Repl Electrode F 35634 40
Repl Electrode F/35634-40
Product Type Accessory
EW-35634-50
In stock.
$185.00 USD/EACH
Qty:
EW-19755-50 YSI Model 3252 Conductivity Cell, Dip Style, ABS plastic, K=1.0
EW-19755-50
Usually ships in 10 days or sooner.
$290.00 USD/EACH
Qty:
EW-19704-70 Thermo Scientific<small><sup>&reg;</sup></small> Orion 4-Cell Conductivity Probe, K = 0.475, 5-ft cable
Thermo Scientific Orion 4-Cell Conductivity Probe, K = 0.475, 5-ft cable
Product Type Accessory,Model 013005MD,Brand Thermo Scientific
EW-19704-70
In stock.
$508.40 USD/EACH
Qty:
760 results (showing 1 - 15 )
Per Page 
Home  >  pH/Ion Meter Accessories
View products by: Title | Brand | Category | Price | Popularity | Best Sellers
View product categories by: Popularity | Our Choices | All-Around Favorites | Title | Product Category | Recommended
View popular items by: Popularity | Favorites | Title | Price | Part Number | Category
View Popular Categories by: Popularity | Our Choices | All-Around Favorites | Product Category | Recommended | Title