United States
[Change]
Shopping Cart
Subtotal
=
$0.00
Items
=
0
View Cart
United States
1-800-323-4340
or
Live Chat
Welcome,
log in
or
register
Search
Shop All Products
Balances & Scales
Baths and Circulators
Calibration
Chemicals, Reagents, & Lubricants
Clean Room
Cooling Equipment
Fittings
Flow, Level & Valves
Fluid & Liquid Handling
Furniture & Fume Hoods
Heating Equipment
Industrial Equipment
Lab Equipment
Laboratory Cleaning Supplies
Laboratory Supplies & Consumables
Life Science
Masterflex Peristaltic Pumps
Masterflex Pump Tubing
Material Handling and Storage
Material Testing & Physical Testing
Mixers and Blenders
pH Meters and Water Quality Products
Pressure & Vacuum Instruments
Process Equipment & Instrumentation
Pumps
Recorders & Data Acquisition
Safety Products
Shop by Industry
Spectroscopy and Spectrophotometers
Temperature, Humidity, Moisture and Weather Products
Tubing, and Hose
Shop by
|
Service & Support
|
Technical Resources
|
My Account
Check order status
Category:
pH Meters, Ion Meters and ISFET Meters
(216)
pH/Ion Buffers, and Storage Solutions
(104)
Brand:
Oakton
(121)
Thermo Scientific Orion
(85)
Cole-Parmer
(30)
[+]
Show more
(9)
Hach
(20)
WTW
(18)
Horiba
(9)
FISHER
(7)
Extech
(5)
Metrepak
(4)
YSI
(3)
Knick
(3)
Palintest
(1)
[-]
Show less
Product Type:
pH Meter
(122)
pH Buffers
(74)
Multiparameter Meter
(56)
[+]
Show more
(17)
pH Tester
(8)
Conductivity Meter
(4)
Vials
(4)
Buffer
(4)
Accessory
(3)
Storage Solutions
(3)
Dissolved Oxygen Meter
(3)
Ion Tester
(3)
ISFET Tester
(3)
Cleaning Solutions
(2)
ORP Standards
(2)
Reference Fill Solutions
(2)
ORP Tester
(2)
pH Module
(2)
Accessory, Probe
(2)
Calibration Solutions
(1)
Ion Meters
(1)
[-]
Show less
Price Range:
$0 to $100
(98)
$100 to $299
(34)
$300 to $499
(31)
[+]
Show more
(4)
$500 to $799
(59)
$800 to $1,099
(36)
$1,100 to $1,999
(46)
$2,000 and Over
(16)
[-]
Show less
Availability:
In Stock
(149)
CE Compliance:
Yes
(183)
Temperature compensation:
Automatic or manual
(149)
Automatic
(38)
Manual
(5)
Waterproof:
Yes
(133)
Meter style:
Accessory
(2)
Benchtop
(72)
Portable
(112)
[+]
Show more
(1)
Tester
(18)
[-]
Show less
Interface:
USB or RS232
(46)
RS-232
(35)
USB
(15)
[+]
Show more
(1)
Bluetooth
(3)
[-]
Show less
Datalogging (points):
2000
(34)
500
(26)
100
(22)
[+]
Show more
(9)
50
(16)
5000
(14)
200
(7)
16
(5)
999
(4)
1000
(3)
15
(3)
300
(2)
25
(1)
[-]
Show less
Volume (mL):
500
(22)
4000
(12)
475
(10)
[+]
Show more
(3)
60
(10)
1000
(9)
125
(3)
[-]
Show less
Learn more:
Isolating Genomic DNA from Whole Blood
Tips for pH Buffer Use
Oakton?« pH meter becomes a TV star!
ChemicalsSpecial Ordering Information
[+]
Show More
Understanding Low Conductivity/High Resistivity Measurements
[-]
Show Less
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here.
Call
1-800-323-4340
Option
9
ext.
6334
See Details
Home
>
Shop by Industry
> pH Measurement
pH Measurement
Cole-Parmer offers an extensive selection of pH meters, and pH instruments for testing and measuring pH
pH measurement is one of the most important measurements taken in science, industrial, and food processing applications. What is pH? In Chemistry, pH is the measurement of the acidity or basicity of an aqueous solution or food products. Pure water is considered neutral, with a pH value of 7.0. A pH value less than 7.0 is considered acidic and pH greater than 7.0 is considered basic or alkaline.
pH measurements have critical implications in a wide variety of applications and disciplines including biology, chemistry, food science, engineering, environmental testing, medicine, and much more.
Whether you are checking the pH of solutions and products in your biology or chemistry lab, or checking the pH values of lakes and streams, or you want to evaluate the pH levels of your household swimming pool or fish aquarium, our selection of pH meters, pH buffers, and pH testing accessories is among the best.
Close
Save My Shopping List
Please login in order to use Shopping Lists
Close
Close
Save My Shopping List
You can
view your shopping list
or
continue shopping
Compare
Product Detail
Sort
Most Popular
Name
Name Descending
Price (Lowest First)
Price (Highest First)
Availability
Price
Oakton Waterproof pHTestr 30 Pocket pH Tester
Meter style Tester,Dimensions " L 6 1/2,Dimensions " Dia 1 1/2
EW-35634-30
In stock.
$116.00 USD/EACH
Qty:
Add to Cart
Save to shopping list
Accessories
Calibrations
More Details
See all
Oakton
Waterproof pHTestr
30 Pocket pH Tester
Oakton Waterproof EcoTestr
pH 2 Pocket pH Tester
Product Type pH Tester,Meter style Tester,Dimensions " W 1-5/8
EW-35423-10
In stock.
$62.00 USD/EACH
Qty:
Add to Cart
Save to shopping list
Accessories
Calibrations
More Details
See all
Oakton
Waterproof EcoTestr
pH 2 Pocket pH Tester
Oakton Waterproof pHTestr 10 Pocket pH Tester
Product Type pH Tester,Meter style Tester,Dimensions " L 6-1/2
EW-35634-10
In stock.
$96.00 USD/EACH
Qty:
Add to Cart
Save to shopping list
Accessories
Calibrations
More Details
See all
Oakton Waterproof pHTestr 10 Pocket pH Tester
Oakton pH 700 Benchtop Meter, double-junction refillable glass pH electrode, ATC probe and stand
Product Type pH Meter,Meter style Benchtop,Dimensions " W 6-7/8
EW-35419-10
In stock.
$631.00 USD/EACH
Qty:
Add to Cart
Save to shopping list
Accessories
Calibrations
More Details
See all
Oakton pH 11 meter with "All-in-One" electrode (35811-71)
Product Type pH Meter,Meter style Portable,Dimensions " W 3-1/2
EW-35614-20
In stock.
$362.00 USD/EACH
Qty:
Add to Cart
Save to shopping list
Accessories
Calibrations
More Details
See all
Oakton
pH 11 Handheld pH Meters
Oakton pH 5+ meter with probe
Product Type pH Meter,Meter style Portable,Dimensions " W 3-3/8
EW-35613-52
In stock.
$263.00 USD/EACH
Qty:
Add to Cart
Save to shopping list
Accessories
Calibrations
More Details
See all
Oakton
pH 5+ Handheld pH Meters
Oakton Waterproof pHTestr 20 (-1.00 to 15.00 pH Range)
Product Type pH Tester,Meter style Tester,Dimensions " L 6-1/2
EW-35634-20
In stock.
$105.00 USD/EACH
Qty:
Add to Cart
Save to shopping list
Accessories
Calibrations
More Details
See all
Oakton Waterproof pHTestr 20 Pocket pH Tester
Oakton pH 11 meter kit
Product Type pH Meter,Meter style Portable,Dimensions " W 3-1/2
EW-35614-80
In stock.
$423.00 USD/EACH
Qty:
Add to Cart
Save to shopping list
Accessories
Calibrations
More Details
See all
Oakton
pH 11 Handheld pH Meters
Oakton pH 700 Benchtop Meter with "all-in-one" pH electrode
Product Type pH Meter,Meter style Benchtop,Dimensions " W 6-7/8
EW-35419-03
In stock.
$508.00 USD/EACH
Qty:
Add to Cart
Save to shopping list
Accessories
Calibrations
More Details
See all
Oakton pH meter with probe
Product Type pH Meter,Meter style Portable,Dimensions " W 3-3/8
EW-35613-22
In stock.
$290.00 USD/EACH
Qty:
Add to Cart
Save to shopping list
Accessories
Calibrations
More Details
See all
Oakton
pH 6+ Handheld pH Meters
Oakton 7.00 pH buffer, 1 L
Product Type pH Buffers,Description pH 7.00,Volume mL 1000
EW-05942-42
In stock.
$20.70 USD/EACH
Qty:
Add to Cart
Save to shopping list
Accessories
Calibrations
More Details
See all
Oakton pH Buffer Solutions
Oakton 7.00 pH buffer, 500-mL
Product Type pH Buffers,Description pH 7.00,Volume mL 500
EW-00654-04
In stock.
$13.35 USD/PINT
Qty:
Add to Cart
Save to shopping list
Accessories
Calibrations
More Details
See all
Oakton pH Buffer Solutions
Oakton 7.00 pH buffer, 4 L
Product Type pH Buffers,Description pH 7.00,Volume mL 4000
EW-05942-44
In stock.
$42.00 USD/EACH
Qty:
Add to Cart
Save to shopping list
Accessories
Calibrations
More Details
See all
Oakton pH Buffer Solutions
Oakton pH 300 meter with single-junction pH/ATC probe
Product Type pH Meter,Meter style Portable,Dimensions " W 3-1/2
EW-35618-03
In stock.
$572.00 USD/EACH
Qty:
Add to Cart
Save to shopping list
Accessories
Calibrations
More Details
See all
Oakton
Waterproof pH 300 pH Meters
Oakton 4.01 pH buffer, 1 L
Product Type pH Buffers,Description pH 4.01,Volume mL 1000
EW-05942-22
In stock.
$20.70 USD/EACH
Qty:
Add to Cart
Save to shopping list
Accessories
Calibrations
More Details
See all
Oakton pH Buffer Solutions
320
results (showing
1
-
15
)
1
2
3
4
5
6
Next >
>>
Per Page
5
10
15
25
50
Home
>
Shop by Industry
> pH Measurement
View products by:
Title
|
Brand
|
Category
|
Price
|
Popularity
|
Best Sellers
View product categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Title
|
Product Category
|
Recommended
View popular items by:
Popularity
|
Favorites
|
Title
|
Price
|
Part Number
|
Category
View Popular Categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Product Category
|
Recommended
|
Title