pHydrion Paper pH Strips
pHydrion Paper pH Strips
  • Replaces two vials of old style litmus
These low-cost disposable vials of pHydrion pH paper strips test pH ranging from 0 to 12. Box of ten vials; 100 strips per vial.
 Product Detail
Sort
 Availability
 Price
EW-59200-25 pHydrion 1-12 Paper Strips
pHydrion 1-12 Paper Strips
Description pHydrion 1-12 Paper Strips,Model 165/1-12
EW-59200-25
In stock.
$15.10 USD/CARTON of 10
Qty:
1 results (showing 1 - 1 )
Per Page 
View products by: Title | Brand | Category | Price | Popularity | Best Sellers
View product categories by: Popularity | Our Choices | All-Around Favorites | Title | Product Category | Recommended
View popular items by: Popularity | Favorites | Title | Price | Part Number | Category
View Popular Categories by: Popularity | Our Choices | All-Around Favorites | Product Category | Recommended | Title