Product Info

Product Info        
Brand Thermo Scientific Orion Sensorex
Item Model PH1-293 None
Product Type pH Meter pH Meter
Unit Of Measure Each Each
Number of Items 1 1
Warranty Period Unit 1 Year 1 Year
Product Name PH-1 Module for iPhone/iPad/iPod SAM-1 (Smart Aqua Meter) Multiparameter Module for Smartphones or Tablets

Technical Specs

Technical Specs        
Meter Type Portable Portable
Connections BNC
Min pH (pH) 0 0
Max pH (pH) 14 14
pH Resolution 0.01/0.1 0.01
pH Accuracy ±0.01 pH ±0.01
Buffer Recognition NIST and USA
Calibration Up to 5 points
Min mV (mV) -1250 -1250
Max mV (mV) 1250 1250
mV Resolution 1 or 10 mV 1 mV
mV Accuracy ±1mV 1 digit ±1
Min Temperature (° C) 0
Max Temperature (° C) 100
Min Temperature (° F) 32
Max Temperature (° F) 212
Warranty 1 year 1 year
Min Conductivity 0 to 5000 µS
Conductivity Accuracy ±5% FS
Temperature compensation Automatic or manual

Compliance

Compliance        
Agency Compliance CE