Product Info

Product Info        
Brand Horiba Oakton
Item Model 352734-1
Product Type Multiparameter Meter Multiparameter Meter
Unit Of Measure Each Each
Warranty Period Unit 1 Year 3 Years
Product Name Horiba D-54 pH/Conductivity Meter Oakton® Waterproof pH/Con 300 pH/Conductivity Meters

Technical Specs

Technical Specs        
Meter Type Portable Portable
Display type LCD LCD
Min pH (pH) 0 -2
Max pH (pH) 14 16
pH Resolution 0.01/0.1 0.01/0.1
pH Accuracy ±0.01 pH ±0.01 pH
Buffer Recognition DIN, NIST, and USA NIST and USA
Min mV (mV) -1999
Max mV (mV) 1999
mV Resolution 1 mV
mV Accuracy ±1mV 1 digit
Conductivity Accuracy ±0.5% FS 1 LSD ±1% FS
Cell Constant Adjustable
Temperature Accuracy ±0.1°C ±1 digit
Temperature compensation Automatic Automatic or manual
Waterproof Yes Yes
Warranty 1 year 3 year
Calibration Up to 5 points
Min Conductivity 0 to 199.9 mS
Conductivity Resolution 0.01 or 0.1 μS; 0.001, 0.01, or 0.1 mS
Min TDS 0 to 200 ppt
TDS Resolution 0.01 or 0.1 ppm; 0.001, 0.01, or 0.1 ppt
TDS Accuracy ±1% of reading

Compliance

Compliance        
Agency Compliance Prop 65 NA (no Prop 65 chemicals) CE