Product Info
Product Info        
Brand Thermo Scientific Nicolet Thermo Scientific Nicolet
Item Model 912A0712
Product Type Infrared Spectrophotometer Infrared Spectrophotometer
Unit Of Measure Each Each
Number of Items 1
Warranty Period Unit 1 Year 1 Year
Product Name Thermo Scientific NICOLET IR100 FT-IR Spectrometer Thermo Scientific Nicolet iS5 FT-IR Spectrometer
Technical Specs
Technical Specs        
Source Lamp Mid-IR ceramic emitter Mid-IR ceramic emitter
Detector Deuteriated tri-glyceride sulfate (dTGS) Deuteriated tri-glyceride sulfate (dTGS)
Scan Time <1 second
Output Ethernet USB
Display Type LCD
Warranty 1 year 1 year
Min Spectral (cm-¹) 350
Max Spectral (cm-¹) 7800
Compliance
Compliance        
Agency Compliance CE;cETLus