Product Info

Product Info        
Brand Fisher Chemical Cole-Parmer
Item Model SS2904 None
Product Type Chemical Reagent Inorganic Chemical
Unit Of Measure Each Each
Number of Items 1
Warranty Period Unit Out Of Box None
Product Name Fisher Chemical™ Sodium Hypochlorite Solution Cole-Parmer® Sodium Hypochlorite Solution

Technical Specs

Technical Specs        
Volume (mL) 4000 4000
CAS Number 7681-52-9 7681-52-9
Form Liquid
Packaging Block Poly Bottle

Compliance

Compliance        
Agency Compliance Prop 65 NA (no Prop 65 chemicals)