Product Info

Product Info        
Brand Acros Organics Cole-Parmer
Item Model None None
Product Type Chemical Reagent Inorganic Chemical
Unit Of Measure Each Each
Number of Items 1 1
Warranty Period Unit Out Of Box None
Product Name Acros Organics Potassium Sulfate Cole-Parmer® Potassium Sulfate, ACS Grade

Technical Specs

Technical Specs        
Weight (g) 500 500
CAS Number 7778-80-5 7778-80-5
Purity Grade ACS Reagent
Class Reagent Salts
Form Solid
Packaging Poly Bottle

Compliance

Compliance        
Agency Compliance Prop 65 NA (no Prop 65 chemicals)