Product Info

Product Info        
Brand Acros Organics Cole-Parmer
Item Model None None
Product Type Chemical Reagent Inorganic Chemical
Unit Of Measure Each Each
Number of Items 1 1
Warranty Period Unit Out Of Box None
Product Name Acros Organics Sodium Chlorite, 80% Cole-Parmer® Sodium Chlorite, Technical Grade

Technical Specs

Technical Specs        
Weight (g) 500 500
CAS Number 7758-19-2 7758-19-2
Purity Grade Technical
Class Reagent Salts
Form Solid
Packaging Poly Bottle

Compliance

Compliance        
Agency Compliance Prop 65 NA (no Prop 65 chemicals)