Subtotal = $0.00 
Items = 0 
View Cart
United States
1-800-323-4340 or 
Welcome,
log in or register
Custom Signs
A sign needs to be quickly seen and easily understood to make the maximum impact. You know exactly what message needs to be conveyed by the signs in your facility. Now you can design your own customized signs by following these easy steps:
1. Choose the sign material that best meets your application.
  • Adhesive Vinyl—this flexible, 4-mil pressure-sensitive material is ideal for your low-cost, lightduty applications.
  • Plastic—this medium-duty, general-purpose, 0.055"-thick polyethylene material is perfect for indoor and sheltered outdoor locations.
  • Aluminum—this 0.040"-thick material offers the strength of metal but without rusting. The surface is overlaminated to protect from splashes and dirt for long-term outdoor exposure.
2. Select OSHA or ANSI header if required to designate the severity of the hazard.
  • Danger (red)
  • Caution (yellow)
  • Warning (orange)
  • Notice (blue)
  • Safety First (green)
See Construction of a Safety Sign (pdf) for definitions of these hazards.
3. Select from the industry standard sizes below or tell us what special size you need.
4. We will work together to design a sign that best meets your unique needs.
  • Simply write or draw a concept, or use our easy-to-follow custom sign planning form on coleparmer.com/custom-signs.
  • Add words, colors, logos, symbols, images, and even photo-quality graphics!
  • We will lay out your message and develop the design, creating your custom sign for maximum effect. State-of-the-art printing produces your sign in the most accurate format.
To create your custom signs with the help of our Customer Service Representatives, please call 800-323-4340 ext. SIGN (7446).
Outside the U.S. contact us
Size
Price per sign (Shown in USD)
1
2 to 5
6 to 9
10+
Adhesive vinyl
5" x 7"
$ 42.00
$ 24.00
$15.00
$ 8.00
7" x 10"
  44.00
  26.00
  16.00
 10.00
10" x 14"
  47.00
  29.00
  20.00
 13.00
14" x 20"
  74.00
  47.00
  33.00
 24.00
20" x 28"
  90.00
  62.00
  48.00
 38.00
Custom size
Call for a quote
Plastic
5" x 7"
  46.00
  27.00
  16.00
  9.00
7" x 10"
  48.00
  29.00
  19.00
12.00
10" x 14"
  53.00
  34.00
  24.00
17.00
14" x 20"
  84.00
  56.00
  41.00
30.00
20" x 28"
111.00
  77.00
  58.00
44.00
Custom size
Call for a quote
Aluminum
5" x 7"
  46.00
  27.00
18.00
12.00
7" x 10"
  47.00
  32.00
23.00
14.00
10" x 14"
  56.00
  41.00
32.00
27.00
14" x 20"
  92.00
  70.00
57.00
48.00
20" x 28"
143.00
111.00
90.00
76.00
Custom size
Call for a quote
Construction of a Safety Sign and Custom Sign Order Form
Complete Line of Safety Signs and Labels
For help from our Customer Service Representatives,
call 800-323-4340 ext. SIGN (7446).
Outside the U.S. contact us
Unique problems require unique products. Let us help you get it right!
View all the FREE Product Applications Support we offer.
View products by: Job Title | Category | Brand | Part Number | Price
View product categories by: Popularity | Favorites | Job Title | Price | Part Number
View popular items by: Popularity | Our Choices | All-Around Favorites | Job Title
View reviews by: Popularity | Product Title | Best Rated