United States
[Change]
Shopping Cart
Subtotal
=
$0.00
Items
=
0
View Cart
United States
1-800-323-4340
or
Live Chat
Welcome,
log in
or
register
Search
Shop All Products
Balances & Scales
Baths and Circulators
Calibration
Chemicals, Reagents, & Lubricants
Clean Room
Cooling Equipment
Flow, Level & Valves
Fluid & Liquid Handling
Furniture & Fume Hoods
Heating Equipment
Industrial Equipment
Lab Equipment
Laboratory Cleaning Supplies
Laboratory Supplies & Consumables
Life Science
Masterflex Peristaltic Pumps
Masterflex Pump Tubing
Material Handling and Storage
Material Testing & Physical Testing
Mixers and Blenders
pH Meters and Water Quality Products
Pressure & Vacuum Instruments
Process Equipment & Instrumentation
Pumps
Recorders & Data Acquisition
Safety Products
Shop by Industry
Spectroscopy and Spectrophotometers
Temperature, Humidity, Moisture and Weather Products
Tubing, Fittings, and Hose
Shop by
|
Service & Support
|
Technical Resources
|
My Account
Check order status
Define your Application Requirements.
What flow rate do I need?
What chemical am I pumping?
Do conditions such as fluid viscosity or back pressure play a role in my application?
Select a Pump
Series Based on
your Flow Rate.
C/L
®
- Compact/Low flow
0.002 to 43 mL/min
L/S
®
- Laboratory Standard
0.0005 to 3400 mL/min
I/P
®
- Industrial Process
0.012 to 26 LPM
B/T
®
- Bulk Transfer
0.3 to 37 LPM
Select a
Pump Head,
Tubing, and Drive.
The L/S and I/P pump
series are divided into three component categories—
pump head, tubing, and
drive. To complete your
tubing pump system,
choose a pump head,
tubing, and drive from
within the same series.
Home
>
Shop by
>
Masterflex® Pump Configurator
Selecting Your Masterflex
®
Tubing Pump System
Masterflex® Pump Configurator
Enter your specific application parameters
into our interactive Pump Configurator
and in seconds it will select a pump system
that will meet your needs.
Enter Here:
Masterflex
®
Pump Configurator
for selecting your
perfect pump click here
Components of a complete system—What to Consider...
Pump Head
Flow rate
—Different pump heads have different flow rate capabilities.
Tubing change frequency
—Some pump heads enable rapid tubing changes.
Desired number of channels
—Pump heads are stackable in most cases.
Chemical compatibility
—If chemicals spill, pump head materials are important.
Accuracy
— Standard pump heads offer the most accurate, repeatable flow rates.
Tubing
Flow rate
— Tubing size directly affects flow rate.
Chemical compatibility
—See
Masterflex® Tubing & Chemical Compatibility
Masterflex® Tubing & Food Compatibility
Masterflex® Roller Compatibility
Tubing life
—See
chart
Fluid viscosity, presence of solids, back pressure, or suction lift
—
High-performance tubing is best for these conditions.
See
Tubing Selection Guide for Viscous Fluids,
Masterflex® Vacuum/Suction Lift Data
Fluid temperature
—Information concerning the temperature range
for each tubing formulation can be found on Virtual Catalog pages 1450–1453.
Drive
Flow rate
—The rpm of the drive directly correlates to flow rate.
Fixed- or variable-speed
—Variable-speed drives enable flow rate adjustment.
Accuracy of flow rate or dispense volume
—Many drives have digital speed
control circuitry for highly accurate and repeatable flow rates.
Remote control or computer interface
—Some drives can connect
to your process controller or can be operated by a foot switch.
Environmental protection against hazards
—Many drives are sealed against
dust and water exposure.
Purchase your Masterflex
L/S or I/P system
by components ...
1. PUMP HEAD
2. TUBING
3. DRIVE
...or purchase a complete Masterflex system with a
single item number:
C/L Tubing Pumps
L/S Tubing Pumps
I/P Tubing Pumps
B/T Tubing Pumps
Masterflex
®
Encyclopedia
Detailed technical information
and complete listings of Masterflex® parts and accessories all in one resource.
Details
View products by:
Title
|
Brand
|
Category
|
Price
|
Popularity
|
Best Sellers
View product categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Title
|
Product Category
|
Recommended
View popular items by:
Popularity
|
Favorites
|
Title
|
Price
|
Part Number
|
Category
View Popular Categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Product Category
|
Recommended
|
Title