|
Cole-Parmer offers a wide selection of digital microscope cameras. A variety of styles are available from economical, stereo and stereozoom camera microscopes, to full-size and modular microscopes with universal and boom stands. In addition, you'll find high quality digital camera microscopes with stepless magnification and stereo microscopes with built-in digital cameras. Check out our catalog's selections and you'll find the digital microscope camera with the features you need, like some of the features of the Model EW-48925-30 (pictured) listed below.
|
See Entire Line... Digital Microscope Camera Quick Link
Tech Tools
|
|
Model EW-48925-30 — Stereozoom Digital Camera Microscopes deliver clear, upright, three-dimensional images at an economical price. A simple USB connection with your PC or Mac lets you put this plug-and-play stereozoom microscope to work. The built-in digital camera features powerful analysis and manipulation software that counts and measures objects. Enhance saved images with audio narration recordings using MIG format. This stereozoom microscope has widefield 10x eyepieces with dual diopter and interpupillary adjustability from 54 to 76 mm. Each digital microscope camera is supplied with both frosted glass and reversible black-and-white stage plates and a vinyl dust cover.
|
|
Our 2009/2010
Cole-Parmer
General Catalog -
features 60,000+ products
Request your FREE copy of our New General Catalog that has become an industry resource today!
|