|
Choose digital multimeters for higher accuracy and advanced features such as relative measurements, temperature readings, and maximum, minimum, and average readings. Most digital multimeters measure voltage, current, resistance, frequency and temperature and perform continuity and diode testing. Check out our catalog's selections and you'll find the digital multimeters with the features you need, like some of the features of the Model EW-26088-10 (pictured) listed below.
Digital Multimeters Quick Link
Model EW-26088-10 — Fluke® 180-series True rms Multimeters. This LCD digital multimeter advanced features include: faster readings, extended ranges, lower DC volts, dual displays, broader capacitance range, and more. The backlit dual display features a 50,000 count digital display and a 51 segment analog bargraph. Optional ToolPakTM accessory allows you to hang your meter with a magnet or strap and the optional FlukeView® Forms software stores and analyzes your reading on your PC and creates custom documentation.
|
Our 2009/2010
Cole-Parmer
General Catalog -
features 60,000+ products
Request your FREE copy of our New General Catalog that has become an industry resource today!
|