Catalog Search Help
Please Login
View Shopping Cart 0 Items
in cart
Checkout
Catalog  |  Table of Contents  |  Shopping Cart  |  Catalog Help
Promotions
Quick Order!
Cole-Parmer Catalog > Digital Pressure Gauges
Digital Pressure Gauges
Whether you need to monitor system pressure or to actuate other system components, Cole-Parmer offers a wide selection of digital pressure gauges. Our new digital pressure gauges provide the accuracy and convenience you need for solid and reliable results. Digital pressure gauges are available in a wide range of models including low cost, stainless steel cases, and process gauges with visual alarms. Check out our catalog's selections and you'll find the digital pressure gauges with the features you need, like some of the features of the Model EW-68338-00 (pictured) listed below.
See Entire Line...
Digital Pressure Gauge
Quick Link
Tech Tools
Model EW-68338-00 — This Digital Industrial Gauge is perfect for pharmaceutical and biotech with its .25% terminal point accuracy. Operate with battery, 4 to 20 mA, or in- line 12 to 36 VDC.
Quarter percent accuracy includes linearity, hysteresis, repeatability, and terminal point accuracy. Socket and sensor are stainless steel and welded together for added durability. Gauge also has the added versatility of ten engineering units, a menu-driven display of additional features, and password- protected calibration with zero and span.
A small sampling of the thousands of other products available from the Cole-Parmer catalog:
Analytical Balances  
Disposable Nitrile Gloves  
Flow Meters  
Digital Multimeters  
Digital Pressure Gauges  
Digital Microscope Cameras  
Stereo Microscopes  
Infrared Thermometers  
Lab Mixers  
Lab Supplies  
Laboratory Fume Hoods  
Nalgene Bottles  
Peristaltic Pump Tubing  
pH Testers  
Plastic Fittings  
Soil pH Meters  
Silicone Tubing  
Digital Pressure Gauge Center
digital pressure gauges
Our 2009/2010
Cole-Parmer
General Catalog -
features 60,000+ products Request your FREE copy of our New General Catalog that has become an industry resource today!