Catalog Search Help
Please Login
View Shopping Cart 0 Items
in cart
Checkout
Catalog  |  Table of Contents  |  Shopping Cart  |  Catalog Help
Promotions
Quick Order!
Cole-Parmer Catalog > Flow Meters
Flow Meters
Whether you need to monitor water or gas flow rates, Cole-Parmer offers a wide selection of flow meters, including variable area, thermal mass, turbine, ultrasonic, paddle-wheel and more. Our new flow meters provide the accuracy and convenience you need for solid and reliable results. Flow meters are available in a wide range of models. Check out our catalog's selections and you'll find the flow meters with the features you need, like some of the features of the Model EW-03229-05 (pictured) listed below.
Flow Meters Quick Link
Model EW-03229-05 — Correlated Flow meters with High-Resolution Valves. The longer 150-mm scale makes these flow meters perfect for applications demanding high resolution. Ideal for low-flow applications or for any application where you need precise flow control with ±2% full-scale accuracy. Each flowmeter consists of a heavy-walled glass flowtube mounted in an anodized aluminum frame with white acrylic backplate (1/8" thick).
All flow meters come with correlation data sheets for water and air at standard temperature and pressure (STP). We can also supply calibration data for oxygen, nitrogen, hydrogen, helium, CO2, argon, other liquids and gases, and for conditions other than STP—call our Application Specialists for more information.
A small sampling of the thousands of other products available from the Cole-Parmer catalog:
Analytical Balances  
Disposable Nitrile Gloves  
Flow Meters  
Digital Multimeters  
Digital Pressure Gauges  
Digital Microscope Cameras  
Stereo Microscopes  
Infrared Thermometers  
Lab Mixers  
Lab Supplies  
Laboratory Fume Hoods  
Nalgene Bottles  
Peristaltic Pump Tubing  
pH Testers  
Plastic Fittings  
Soil pH Meters  
Silicone Tubing  
flow meters
Our 2009/2010
Cole-Parmer
General Catalog -
features 60,000+ products Request your FREE copy of our New General Catalog that has become an industry resource today!