|
Get quick and accurate surface temperature results with infrared thermometers from Cole-Parmer. Choose from non-laser or sighted to highlight your target measuring area. Models available with different distance-to-target ratios, including close focus infrared thermometers, for accurate measurement at various distances. Specialty infrared thermometers designed specifically for glass, plastics, or metals applications are also available. Check out our catalog's selections and you'll find the infrared thermometers with the features you need, like some of the features of the Model EW-35639-20 (pictured) listed below.
Infrared Thermometers Quick Link
Model EW-35639-20 — Oakton? InfraProTM Infrared Thermometers. Rugged enough for industrial use yet compact and lightweight enough to be carried around with you, these infrared thermometers are extremely easy to use. Just aim, pull the trigger, and read the display. The 3-1/2-digit backlit display shows temperature readings in ?F or ?C; low-battery indication; and hold, scan, and max. Display holds for 7 seconds.
|
Our 2009/2010
Cole-Parmer
General Catalog -
features 60,000+ products
Request your FREE copy of our New General Catalog that has become an industry resource today! |