|
Whether your uses encompass pharmaceutical, chemical, or other commercial applications, Cole-Parmer offers a wide selection of peristaltic pump tubing. Our peristaltic pump tubing comes in a variety of specifications to fit your particular needs. Choose from a large variety of precision, sanitary and spooled peristaltic pump tubing. Check out our catalog's selections and you'll find the peristaltic pump tubing with the features you need, like some of the features of the Model EW-96410-73 (pictured) listed below.
Peristaltic Pump Tubing Quick Link
Model EW-96410-73 — Precision Silicone (platinum) Tubing. Platinum cured silicone peristaltic pump tubing provides excellent biocompatability with very low extractability to provide fluid purity. Translucent for easy flow monitoring, it is plasticizer-free and can be sterilized. Masterflex platinum cured silicone pump tubing is manufactured under FDA bulk pharmaceutical cGMP's and 5 and meets FDA, USP Class 6, EP, and 3A standards.
|
Our 2009/2010
Cole-Parmer
General Catalog -
features 60,000+ products
Request your FREE copy of our New General Catalog that has become an industry resource today!
|