Catalog Search Help
Please Login
View Shopping Cart 0 Items
in cart
Checkout
Catalog  |  Table of Contents  |  Shopping Cart  |  Catalog Help
Promotions
Quick Order!
Cole-Parmer Catalog > Silicone Tubing
Silicone Tubing
Whether your uses encompass pharmaceutical, biotechnology, or aerospace applications, Cole-Parmer offers a wide selection of silcone tubing. Our silicone tubing comes in a variety of specifications to fit your particular needs. Choose from a large variety of platinum-cured, peroxide-cured, low absorption, reinforced and color-coded silicone tubing. Check out our catalog's selections and you'll find the silicone tubing with the features you need, like some of the features of the Model EW-95802-05 (pictured) listed below.
Silicone Tubing Quick Link
Model EW-95802-05 — Platinum-Cured Silicone Tubing. Characteristics include excellent heat resistance, low compression set, odorless, nontoxic and translucent. Resin is FDA-compliant (21 CFR 177.2600); meets USP 23 Class VI, USDA. Tubing is manufactured according to GMP. Temperature range from –60 to 460°F (–51 to 238°C).
A small sampling of the thousands of other products available from the Cole-Parmer catalog:
Analytical Balances  
Disposable Nitrile Gloves  
Flow Meters  
Digital Multimeters  
Digital Pressure Gauges  
Digital Microscope Cameras  
Stereo Microscopes  
Infrared Thermometers  
Lab Mixers  
Lab Supplies  
Laboratory Fume Hoods  
Nalgene Bottles  
Peristaltic Pump Tubing  
pH Testers  
Plastic Fittings  
Soil pH Meters  
Silicone Tubing  
silicone tubing
Our 2009/2010
Cole-Parmer
General Catalog -
features 60,000+ products Request your FREE copy of our New General Catalog that has become an industry resource today!