|
Cole-Parmer offers a wide selection of pH meters including soil pH meters. Our soil pH meters allow you to easily test and monitor the pH levels for healthy and productive plants. Take a look at our soil pH kits if you need to test additional parameters such as salinity and conuctivity. Check out our catalog's selections and you'll find the soil pH meters with the features you need, like the features of the Model EW-35614-22 (pictured) listed below.
Soil pH Meters Quick Link
Model EW-35614-22 — OaktonŽ pH/mV/Temperature Basic pH 11 Meters. Basic pH 11 meters feature everything you need for general-purpose pH measurements with not-so-basic benefits. Large, dual-LCD lets you view pH (or mV) and temperature values simultaneously, and troubleshooting data with its decipherable icons. Automatic temperature compensation (ATC) ensures accuracy even in fluctuating temperatures. Functions include temperature offset calibration, HOLD function, ready indicator, and auto-off. Electrode slope display capability allows for quick troubleshooting. Recall up to 50 data sets and calibrate up to five pH points from USA and NIST buffers.
|
Our 2009/2010
Cole-Parmer
General Catalog -
features 60,000+ products
Request your FREE copy of our New General Catalog that has become an industry resource today!
|