|
Cole-Parmer offers a wide selection of stereo microscopes. A variety of styles are available from economical and compact stereo microscopes, to full-size and modular microscopes with universal and boom stands. In addition, you'll find high quality stereozoom microscopes with stepless magnification and stereo microscopes with built-in digital cameras. Check out our catalog's selections and you'll find the stereo microscope with the features you need, like some of the features of the Model EW-03905-10 (pictured) listed below.
|
See Entire Line... Stereo Microscopes Quick Link
Tech Tools
|
|
Model EW- 03905-10 — Economy Compact Dual-Power Stereo microscopes deliver clear, upright, three-dimensional images at an economical price. The compact stand saves bench space and stands 11" tall. Microscopes feature features a 45° inclined swivel head with adjustable diopter and interpupillary controls. Heads rotate 360° for front position viewing. Paired 1x/3x or 2x/4x objectives are mounted in a revolving turret. Paired 10x widefield eyepieces have a 34-mm OD and accept reticles with 22.8-mm OD. Working distance is 75 mm. Built-in incident (top) and transmitted (bottom) light sources on all models allow versatile illumination of any sample. Each stereo microscope is supplied with both frosted glass and reversible black-and-white stage plates and a vinyl dust cover.
|
|
Our 2009/2010
Cole-Parmer
General Catalog -
features 60,000+ products
Request your FREE copy of our New General Catalog that has become an industry resource today!
|