Catalog Search Help
Please Login
View Shopping Cart 0 Items
in cart
Checkout
Catalog  |  Table of Contents  |  Shopping Cart  |  Catalog Help
Promotions
Quick Order!
Cole-Parmer Catalog > Digital Microscope Cameras
Digital Microscope Cameras
Cole-Parmer offers a wide selection of digital microscope cameras. A variety of styles are available from economical, stereo and stereozoom camera microscopes, to full-size and modular microscopes with universal and boom stands. In addition, you'll find high quality digital camera microscopes with stepless magnification and stereo microscopes with built-in digital cameras. Check out our catalog's selections and you'll find the digital microscope camera with the features you need, like some of the features of the Model EW-48925-30 (pictured) listed below.
See Entire Line...
Digital Microscope Camera Quick Link
Tech Tools
Model EW-48925-30 — Stereozoom Digital Camera Microscopes deliver clear, upright, three-dimensional images at an economical price. A simple USB connection with your PC or Mac lets you put this plug-and-play stereozoom microscope to work. The built-in digital camera features powerful analysis and manipulation software that counts and measures objects. Enhance saved images with audio narration recordings using MIG format. This stereozoom microscope has widefield 10x eyepieces with dual diopter and interpupillary adjustability from 54 to 76 mm. Each digital microscope camera is supplied with both frosted glass and reversible black-and-white stage plates and a vinyl dust cover.
A small sampling of the thousands of other
products available from the Cole-Parmer catalog:
Analytical Balances  
Disposable Nitrile Gloves  
Flow Meters  
Digital Multimeters  
Digital Pressure Gauges  
Digital Microscope Cameras  
Stereo Microscopes  
Infrared Thermometers  
Lab Mixers  
Lab Supplies  
Laboratory Fume Hoods  
Nalgene Bottles  
Peristaltic Pump Tubing  
pH Testers  
Plastic Fittings  
Soil pH Meters  
Silicone Tubing  
Digital Pressure Gauge Center
Digital Microscope Camera
Our 2009/2010
Cole-Parmer
General Catalog -
features 60,000+ products Request your FREE copy of our New General Catalog that has become an industry resource today!