Catalog Search Help
Please Login
View Shopping Cart 0 Items
in cart
Checkout
Catalog  |  Table of Contents  |  Shopping Cart  |  Catalog Help
Promotions
Quick Order!
Cole-Parmer Catalog > Digital Multimeters
Digital Multimeters
Choose digital multimeters for higher accuracy and advanced features such as relative measurements, temperature readings, and maximum, minimum, and average readings. Most digital multimeters measure voltage, current, resistance, frequency and temperature and perform continuity and diode testing. Check out our catalog's selections and you'll find the digital multimeters with the features you need, like some of the features of the Model EW-26088-10 (pictured) listed below.
Digital Multimeters Quick Link
Model EW-26088-10 — Fluke® 180-series True rms Multimeters. This LCD digital multimeter advanced features include: faster readings, extended ranges, lower DC volts, dual displays, broader capacitance range, and more. The backlit dual display features a 50,000 count digital display and a 51 segment analog bargraph. Optional ToolPakTM accessory allows you to hang your meter with a magnet or strap and the optional FlukeView® Forms software stores and analyzes your reading on your PC and creates custom documentation.
A small sampling of the thousands of other products available from the Cole-Parmer catalog:
Analytical Balances  
Disposable Nitrile Gloves  
Flow Meters  
Digital Multimeters  
Digital Pressure Gauges  
Digital Microscope Cameras  
Stereo Microscopes  
Infrared Thermometers  
Lab Mixers  
Lab Supplies  
Laboratory Fume Hoods  
Nalgene Bottles  
Peristaltic Pump Tubing  
pH Testers  
Plastic Fittings  
Soil pH Meters  
Silicone Tubing  
digital multimeters
Our 2009/2010
Cole-Parmer
General Catalog -
features 60,000+ products Request your FREE copy of our New General Catalog that has become an industry resource today!