|
Whether your uses encompass pharmaceutical, biotechnology, or aerospace applications, Cole-Parmer offers a wide selection of silcone tubing. Our silicone tubing comes in a variety of specifications to fit your particular needs. Choose from a large variety of platinum-cured, peroxide-cured, low absorption, reinforced and color-coded silicone tubing. Check out our catalog's selections and you'll find the silicone tubing with the features you need, like some of the features of the Model EW-95802-05 (pictured) listed below.
Silicone Tubing Quick Link
Model EW-95802-05 — Platinum-Cured Silicone Tubing. Characteristics include excellent heat resistance, low compression set, odorless, nontoxic and translucent. Resin is FDA-compliant (21 CFR 177.2600); meets USP 23 Class VI, USDA. Tubing is manufactured according to GMP.
Temperature range from –60 to 460°F (–51 to 238°C).
|
Our 2009/2010
Cole-Parmer
General Catalog -
features 60,000+ products
Reserve your FREE copy of our New General Catalog that has become an industry resource today!
|