Catalog Search Help
Please Login
View Shopping Cart 0 Items
in cart
Checkout
Catalog  |  Table of Contents  |  Shopping Cart  |  Catalog Help
Promotions
Quick Order!
Cole-Parmer Catalog > Stereo Microscopes
Stereo Microscopes
Cole-Parmer offers a wide selection of stereo microscopes. A variety of styles are available from economical and compact stereo microscopes, to full-size and modular microscopes with universal and boom stands. In addition, you'll find high quality stereozoom microscopes with stepless magnification and stereo microscopes with built-in digital cameras. Check out our catalog's selections and you'll find the stereo microscope with the features you need, like some of the features of the Model EW-03905-10 (pictured) listed below.
See Entire Line...
Stereo Microscopes Quick Link
Tech Tools
Model EW- 03905-10 — Economy Compact Dual-Power Stereo microscopes deliver clear, upright, three-dimensional images at an economical price. The compact stand saves bench space and stands 11" tall. Microscopes feature features a 45° inclined swivel head with adjustable diopter and interpupillary controls. Heads rotate 360° for front position viewing. Paired 1x/3x or 2x/4x objectives are mounted in a revolving turret. Paired 10x widefield eyepieces have a 34-mm OD and accept reticles with 22.8-mm OD. Working distance is 75 mm. Built-in incident (top) and transmitted (bottom) light sources on all models allow versatile illumination of any sample. Each stereo microscope is supplied with both frosted glass and reversible black-and-white stage plates and a vinyl dust cover.
A small sampling of the thousands of other products available from the Cole-Parmer catalog:
Analytical Balances  
Disposable Nitrile Gloves  
Flow Meters  
Digital Multimeters  
Digital Pressure Gauges  
Digital Microscope Cameras  
Stereo Microscopes  
Infrared Thermometers  
Lab Mixers  
Lab Supplies  
Laboratory Fume Hoods  
Nalgene Bottles  
Peristaltic Pump Tubing  
pH Testers  
Plastic Fittings  
Soil pH Meters  
Silicone Tubing  
Digital Pressure Gauge Center
stereo microscopes
Our 2009/2010
Cole-Parmer
General Catalog -
features 60,000+ products Request your FREE copy of our New General Catalog that has become an industry resource today!