Item Total = $0.00 
Items = 0 
View Cart
United States
1-800-323-4340 or 
Welcome,
log in or register
0 to 5 psig Cole-Parmer ±0.25%-Accuracy Gauge Transmitter, 4 to 20 mA Output
Item# YO-68075-10
$159.00 USD/EACH
Are you a reseller or buying in bulk? Call us today.
Wetted materials 17-4 PH SS
Output 4 to 20 mA
Range 0 to 5 psi
Process connection 1/4" NPT(M)
Electrical connections 2-ft (0.6-m) cable
Power 9 to 30 VDC
Accuracy ±0.25% full-scale
CE Compliance Yes
Product Type Pressure Transducer
Brand Cole-Parmer
Manufacturer number 2091005PG2M1102CP1
Qty/ea 1
Calibrations
Qty/Price
Availability
at $155.00 USD/EACH
Usually ships in 7 days.
Technical Resources
Virtual Catalog (PDFs)
  • Catalog - Spanish
  • Catalog - English
  •  
    Service & Support
    Still have questions
    Contact Our Experts
    for FREE product
    application selection
    and support.
    or call 1-800-323-4340
    Custom Ordering Solutions
    Custom Ordering Solutions
    Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here. Call 1-800-323-4340 Option 9 ext. 6334
    Questions and Answers
    Do you have questions about this product?
    get answers from real customers and in-house experts with AnswerBox.
    6 Questions | 7 Answers
    Displaying questions 1-6
    • Q:
      How can I connect this sensor to module Analog of PLC premium if I want signal output 0-5V ?
      Asked on 10/31/2013 by hoangdtbk from hcm
      1 answer
      • STAFF EXPERT
        A:
        Usually PLCs use a 4-20 mA signal, which this unit has.
        Please see part number 68075-40 for the 0.5-5.5 VDC output version of this sensor, if DC output is needed instead.
        Answered on 10/31/2013 by Jen from Technical Support
    • Q:
      What is the internal volume of this pressure transducer?
      Asked on 7/28/2011 by Anonymous
      2 answers
      • STAFF EXPERT
        A:
        The internal volume of this transducer is 1.434 mL.
        Answered on 8/1/2011 by Jack from Technical Support
      • STAFF EXPERT
        A:
        The internal volume of this transducer is 1.434 mL.
        Answered on 8/1/2011 by Jack from Technical Support
    • Q:
      can we use there transducers in wastewater applications. (Membrane bioreactor applications to measure the pressure inside of pipes which has wastewater flow).
      Asked on 1/13/2011 by Amila from AIT, bangkok, Thailand
      1 answer
      • STAFF EXPERT
        A:
        The wetted material for pressure transducer is 17-4 PH stainless steel. As long as your wastewater does not contain materials that will react to the steel this will work in your application.
        Answered on 1/17/2011 by Mike from Tech Support
    • Q:
      Can I connect this pressure transducer with "Current and DC voltage logger"(EJ-23030-30) directly? And store data in the logger and download to a notebook in daily or weekly basis.
      Asked on 1/10/2011 by Amila from AIT, Bangkok, Thailand
      1 answer
      • Staff Reviewer
        A:
        Yes, the 23030-30 datalogger would also be appropriate.
        Answered on 1/13/2011 by Jen from Technical Support
    • Q:
      How can I connect with a data logger. Any suggestions of data loggers to connect with this pressure transducer. We like to connect and store data for one week and then transfer to a notebook through a memory.
      Asked on 1/10/2011 by Amila from AIT, bangkok, Thailand
      1 answer
      • Staff Reviewer
        A:
        There are dataloggers that accept 4-20 mA inputs. Please see something like part number 23039-56.
        Answered on 1/13/2011 by Jen from Technical Support
    • Q:
      This product is rated 0-5 PSIG fot the range of the 4-20ma output. What is the maximum pressure that can be applied without damaging the pressure transducer? ie) if I temporarily apply X PSI above 5 PSIG during a machine cycle will the sensor be damaged?
      Asked on 10/12/2010 by Kedgamatic from OP, NY
      1 answer
      • STAFF EXPERT
        A:
        pressures of 10psi will cause the zero to shift up to +/- 0.5% of the full span. If it sees this happen frequently the diaphragm will eventually yield and fail.
        Burst pressure of the unit 250psi but the unit will have failed long before that.
        Answered on 10/14/2010 by Mike from Tech Support
    Displaying questions 1-6
    Do you have a question about this product? 
    Product Reviews
    Customers Who Bought This Item Also Bought