United States
[Change]
Shopping Cart
Subtotal
=
$0.00
Items
=
0
View Cart
United States
1-800-323-4340
or
Live Chat
Welcome,
log in
or
register
Search
Shop All Products
Balances & Scales
Baths and Circulators
Calibration
Chemicals, Reagents, & Lubricants
Clean Room
Cooling Equipment
Fittings
Flow, Level & Valves
Fluid & Liquid Handling
Furniture & Fume Hoods
Heating Equipment
Industrial Equipment
Lab Equipment
Laboratory Cleaning Supplies
Laboratory Supplies & Consumables
Life Science
Masterflex Peristaltic Pumps
Masterflex Pump Tubing
Material Handling and Storage
Material Testing & Physical Testing
Mixers and Blenders
pH Meters and Water Quality Products
Pressure & Vacuum Instruments
Process Equipment & Instrumentation
Pumps
Recorders & Data Acquisition
Safety Products
Shop by Industry
Spectroscopy and Spectrophotometers
Temperature, Humidity, Moisture and Weather Products
Tubing, and Hose
Shop by
|
Service & Support
|
Technical Resources
|
My Account
Check order status
View Product List
< 5
of
13 >
Representative photo only
12x6x4 Junction Box Carbon Steel
Click to enlarge
More Product Details
Specifications
Accessories
Technical Resources
Related technical articles
Service & Support
Answer Box -Q&A posts
Product Reviews
dummy
Close[X]
EW-30001-20
was succesfully added to your cart.
No Calibration Needed
Yes, Add Calibration
Learn more:
Why calibrate Test Equipment?
Home
>
Furniture & Fume Hoods
>
Junction Boxes
>
Junction Boxes - Carbon Steel (Sizes ranging from: 6x6x4 to 16x14x6)
Share
|
12x6x4 Junction Box, Carbon Steel
EW-30001-20
$66.50 USD/EACH
Usually ships in 7 days.
Qty
:
Add to Cart
Save to shopping list
Provides protection for electrical components for indoor and outdoor environments
Foam-in-place gasket ensures perfect seal
Painted carbon steel RAL 7035 light gray, rated NEMA 12 (IP55), 3R, and 4 (IP66)
Stainless steel units available in 304SS or 316LSS, #4 brushed finish, rated NEMA 4X (IP66)
Galvanized panel included (WM wall-mount only)
Carbon steel enclosures have wall-mounting holes - SS enclosures have wall-mounting blind-nuts
Specifications
Product Type
Junction Box Enclosures
Material
Carbon Steel
Outside dimensions (" W)
12
Outside dimensions (" H)
6
Outside dimensions (" D)
4
Manufacturer number
JB120604HC
Model
JB120604HC
Accessories
Qty
/
Price
Availability
6x6 Jb Mounting Panel
EW-30001-78
at
$5.50 USD/EACH
Usually ships in 7 days.
Add to Cart
8x6 Jb Mounting Panel
EW-30001-79
at
$5.50 USD/EACH
Usually ships in 7 days.
Add to Cart
8x8 Jb Mounting Panel
EW-30001-80
at
$6.50 USD/EACH
Usually ships in 7 days.
Add to Cart
10x8 Jb Mounting Panel
EW-30001-81
at
$7.50 USD/EACH
Usually ships in 7 days.
Add to Cart
10x10 Jb Mounting Panel
EW-30001-82
at
$8.50 USD/EACH
In stock.
Add to Cart
12x10 Jb Mounting Panel
EW-30001-83
at
$10.00 USD/EACH
Usually ships in 7 days.
Add to Cart
12x12 Jb Mounting Panel
EW-30001-84
at
$11.50 USD/EACH
Usually ships in 7 days.
Add to Cart
14x12 Jb Mounting Panel
EW-30001-85
at
$13.00 USD/EACH
Usually ships in 7 days.
Add to Cart
16x14 Jb Mounting Panel
EW-30001-86
at
$16.00 USD/EACH
Usually ships in 7 days.
Add to Cart
JB Wallmount Bracket
EW-30001-87
at
$6.50 USD/PACKAGE
Usually ships in 7 days.
Add to Cart
JB Wallmount Bracket, Type 304 SS
EW-30001-88
at
$29.50 USD/PACKAGE
Usually ships in 7 days.
Add to Cart
JB/WM Thumb Turn Handle, Keylocking
EW-30001-89
at
$21.50 USD/PACKAGE
Usually ships in 7 days.
Add to Cart
JB/WM Thumb Turn Handle
EW-30001-90
at
$56.00 USD/PACKAGE
Usually ships in 7 days.
Add to Cart
WM Wallmount Bracket
EW-30001-92
at
$34.50 USD/PACKAGE
Usually ships in 7 days.
Add to Cart
Technical Resources
Related technical articles (Roll over for summary)
Importance of Airflow Monitors for Fume Hoods
Show Less...
Service & Support
Still have questions?
Contact Our Experts
for FREE product
application selection
and support.
or call 1-800-323-4340
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here.
Call
1-800-323-4340
Option
9
ext.
6334
See Details
Questions and Answers
Product Reviews
Close
Save My Shopping List
Please login in order to use Shopping Lists
Close
Close
Save My Shopping List
You can
view your shopping list
or
continue shopping
View products by:
Title
|
Brand
|
Category
|
Price
|
Popularity
|
Best Sellers
View product categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Title
|
Product Category
|
Recommended
View popular items by:
Popularity
|
Favorites
|
Title
|
Price
|
Part Number
|
Category
View Popular Categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Product Category
|
Recommended
|
Title