log in or register
191-1 : Vibration Transmitter,0-1 in/sec
EW-98853-14
$385.00 USD/EACH
Usually ships in 4 days.
Qty
    FEATURES
    • Solid-state circuitry
    • Two-wire 4-20 mA signal format
    • Same wiring for both power and vibration signal
    • Economical for continuous monitoring of all machines
    • Connects in series with DC power supply and measurement devices
    • Works with single-loop monitors
    • Rugged NEMA 4 weatherproof case
    APPLICATIONS
    • Fans, Motors, Pumps and Compressors
    • Controlling Vibrating Conveyors
    • General Purpose Machinery
    • Hazardous Environments
    Model 191-1
    Calibrations
    Qty/Price
    Availability
    at $155.00 USD/EACH
    Usually ships in 8 days.
    Technical Resources
    Service & Support
    Still have questions?
    Contact Our Experts
    for FREE product
    application selection
    and support.
    or call 1-800-323-4340
    Custom Ordering Solutions
    Custom Ordering Solutions
    Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here. Call 1-800-323-4340 Option 9 ext. 6334
    Questions and Answers
    Product Reviews
    View products by: Title | Brand | Category | Price | Popularity | Best Sellers
    View product categories by: Popularity | Our Choices | All-Around Favorites | Title | Product Category | Recommended
    View popular items by: Popularity | Favorites | Title | Price | Part Number | Category
    View Popular Categories by: Popularity | Our Choices | All-Around Favorites | Product Category | Recommended | Title