United States
[Change]
Shopping Cart
Subtotal
=
$0.00
Items
=
0
View Cart
United States
1-800-323-4340
or
Live Chat
Welcome,
log in
or
register
Search
Shop All Products
Balances & Scales
Baths and Circulators
Calibration
Chemicals, Reagents, & Lubricants
Clean Room
Cooling Equipment
Fittings
Flow, Level & Valves
Fluid & Liquid Handling
Furniture & Fume Hoods
Heating Equipment
Industrial Equipment
Lab Equipment
Laboratory Cleaning Supplies
Laboratory Supplies & Consumables
Life Science
Masterflex Peristaltic Pumps
Masterflex Pump Tubing
Material Handling and Storage
Material Testing & Physical Testing
Mixers and Blenders
pH Meters and Water Quality Products
Pressure & Vacuum Instruments
Process Equipment & Instrumentation
Pumps
Recorders & Data Acquisition
Safety Products
Shop by Industry
Spectroscopy and Spectrophotometers
Temperature, Humidity, Moisture and Weather Products
Tubing, and Hose
Shop by
|
Service & Support
|
Technical Resources
|
My Account
Check order status
View Product List
< 3
of
6 >
Representative photo only
202 5 Inclin Mnmtr 2 0 2inW Durablock Stat Inclined Mnometer 0 2 0 2 0in H2O 8 75in Scle Lngth01SCL Div
Click to enlarge
More Product Details
Specifications
Calibrations
Technical Resources
Virtual Catalog (PDFs)
Related technical articles
Service & Support
Answer Box -Q&A posts
Product Reviews
dummy
Close[X]
EW-68458-99
was succesfully added to your cart.
Would you like it calibrated?
Price
:
Availability
:
EW-17090-31
NIST-Traceable Calibration; Manometers
$140.00 USD/EACH/EACH
Usually ships in 4 days.
No Calibration Needed
Yes, Add Calibration
Learn more:
Why calibrate Test Equipment?
Home
>
Pressure & Vacuum Instruments
>
Digital and Analog Manometers
>
Dwyer Inclined Stationary Manometers
Share
|
202.5:Inclin Mnmtr.2/0/2inW Durablock Stat Inclined Mnometer 0.2/0/2.0in H2O 8.75in Scle Lngth01SCL.Div
EW-68458-99
$494.00 USD/EACH
In stock.
Qty
:
Add to Cart
Save to shopping list
These solid plastic stationary draft gauges are for highly accurate laboratory or general industrial service, for measurement of low range gas and air pressures, positive, negative or differential. Each gauge is furnished with spare gauge oil and rapid shut-off type, molded nylon tubing connectors, two 3' lengths of flexible tubing and 2-1/8" pipe thread adapters.
FEATURES
1" thick acrylic plastic body is a solid block, virtually unbreakable, stable and absolutely free of the danger of distortion
3/16" bores, accurate to ±.0002" are permanently free of bends or crooks, will not require recalibration because of distortion
Ground glass bubble level sensitive to 115/125 Sec/mm is 1/2" long x .375" diameter and provided with two red lines for easy visibility and alignment
Parallax-free reading for maximum accuracy and consistency is achieved by simply aligning the meniscus with its image reflected in the polished aluminum scale
Selected gauge oil with high wettability characteristics forms a consistent well shaped meniscus for perfect readability
O-ring sealed fluid level plunger adjustment for easy zeroing
Specifications
Product Type
Manometers, inclined stationary
Media compatibility
air
Accuracy
±2% full-scale
Range
0.2 to 0 to 2" H
2
O
Process connection
3/16" barb
Liquid fill
0.826 red gauge oil
Brand
Dwyer
Manufacturer number
202.5
Model
202 5
Calibrations
Qty
/
Price
Availability
NIST-Traceable Calibration; Manometers
EW-17090-31
at $140.00 USD/EACH
Usually ships in 4 days.
Add to Cart
Technical Resources
Virtual Catalog (PDFs)
13-14 Catalog
Related technical articles (Roll over for summary)
Anemometers
Absolute, Gauge, and Differential Pressure Scales
Common Pressure Terms
Show All...
Pressure Unit Conversion Constants
The Hidden Costs of Mercury-Filled Vacuum Gauges
Show Less...
Service & Support
Still have questions?
Contact Our Experts
for FREE product
application selection
and support.
or call 1-800-323-4340
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here.
Call
1-800-323-4340
Option
9
ext.
6334
See Details
Questions and Answers
Product Reviews
Close
Save My Shopping List
Please login in order to use Shopping Lists
Close
Close
Save My Shopping List
You can
view your shopping list
or
continue shopping
View products by:
Title
|
Brand
|
Category
|
Price
|
Popularity
|
Best Sellers
View product categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Title
|
Product Category
|
Recommended
View popular items by:
Popularity
|
Favorites
|
Title
|
Price
|
Part Number
|
Category
View Popular Categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Product Category
|
Recommended
|
Title