United States
[Change]
Shopping Cart
Subtotal
=
$0.00
Items
=
0
View Cart
United States
1-800-323-4340
or
Live Chat
Welcome,
log in
or
register
Search
Shop All Products
Balances & Scales
Baths and Circulators
Calibration
Chemicals, Reagents, & Lubricants
Clean Room
Cooling Equipment
Fittings
Flow, Level & Valves
Fluid & Liquid Handling
Furniture & Fume Hoods
Heating Equipment
Industrial Equipment
Lab Equipment
Laboratory Cleaning Supplies
Laboratory Supplies & Consumables
Life Science
Masterflex Peristaltic Pumps
Masterflex Pump Tubing
Material Handling and Storage
Material Testing & Physical Testing
Mixers and Blenders
pH Meters and Water Quality Products
Pressure & Vacuum Instruments
Process Equipment & Instrumentation
Pumps
Recorders & Data Acquisition
Safety Products
Shop by Industry
Spectroscopy and Spectrophotometers
Temperature, Humidity, Moisture and Weather Products
Tubing, and Hose
Shop by
|
Service & Support
|
Technical Resources
|
My Account
Check order status
View Product List
1
of
8 >
EW-81853-01
24/Seven Modular CFR Mat, Solid 3 ft x 3 ft
Click to enlarge
More Product Details
Specifications
Service & Support
Answer Box -Q&A posts
Product Reviews
dummy
Close[X]
EW-81853-01
was succesfully added to your cart.
No Calibration Needed
Yes, Add Calibration
Learn more:
Why calibrate Test Equipment?
Home
>
Safety Products
>
Floor Mats and Matting
>
Wearwell Mats
>
Wearwell 24/Seven Safety Mats
Share
|
24/Seven Modular CFR Mat, Solid 3 ft x 3 ft
EW-81853-01
$103.00 USD/EACH
Usually ships in 3 days.
Qty
:
Add to Cart
Save to shopping list
Modular anti fatique, anti slip mats keep employees safe and comfortable
Grit Works
®
mats should be used where slip hazards such as grease and oil are present.
Customize matting to your desired size using these 3 ft x 3 ft interlocking modules. The CFR rubber mats are for use in areas with mineral oil based cutting fluids and solvents. The nitrile mats are for use in areas with petrochemicals such as hydrocarbons, naphthenes, paraffins, and petroleum distillates. Choose the solid design for dry applications or the open design when drainage is needed. Select the GritWorks surface where slip hazards such as grease and oil are present.
The hidden interlocking system is designed to connect on all four sides to quickly cover large areas. Yellow beveled edging provides protection from tripping and a clean finished look; order separately in table. The female edging snaps onto the protruding mat knobs; the male snaps into the underside of the mat. Each e
Specifications
Product Type
Floor Mats
Thickness
0.625 in (1.6 cm)
Size
3 ft x 3 ft
Color
Black
Material
CFR Rubber
Brand
Wearwell
Manufacturer number
570 58x3x3CFRBK
Model
570 58x3x3CFRBK
Service & Support
Still have questions?
Contact Our Experts
for FREE product
application selection
and support.
or call 1-800-323-4340
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here.
Call
1-800-323-4340
Option
9
ext.
6334
See Details
Questions and Answers
Product Reviews
Close
Save My Shopping List
Please login in order to use Shopping Lists
Close
Close
Save My Shopping List
You can
view your shopping list
or
continue shopping
View products by:
Title
|
Brand
|
Category
|
Price
|
Popularity
|
Best Sellers
View product categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Title
|
Product Category
|
Recommended
View popular items by:
Popularity
|
Favorites
|
Title
|
Price
|
Part Number
|
Category
View Popular Categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Product Category
|
Recommended
|
Title