log in or register
250 ml Allihn condenser
EW-99037-28
$144.00 USD/EACH
Usually ships in 12 days or sooner.
Qty
  • Improved design prevents the trapping of liquid
Use these Soxhlet extraction components to extract soluble components from a solid sample into an organic solvent. Place a sample in the thimble and heat the solvent contained in the flask. As the solvent vapors rise (passing through the extraction thimble), they enter the water-cooled condensor, and liquify. When the liquid level in the extractor reaches the top of the siphon tube, siphoning action returns the extract-enriched solvent to the flask.
Soxhlet-type extraction is used for continuous extraction of solids with a suitable solvent. The improved design includes a protected siphon tube, an enlarged vapor tube, and sloping seals to prevent the trapping of liquid. TS joints connect the condenser to the extractor and the extractor to the flask. Uses paper, alundum, or extraction thimbles (order separately). Purchase Soxhlet glassware as a complete assembly or as individual components.<br>
Joint size 45/50
Technical Resources
Virtual Catalog (PDFs)
  • 13-14 Catalog
  •  
    Service & Support
    Still have questions?
    Contact Our Experts
    for FREE product
    application selection
    and support.
    or call 1-800-323-4340
    Custom Ordering Solutions
    Custom Ordering Solutions
    Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here. Call 1-800-323-4340 Option 9 ext. 6334
    Questions and Answers
    Product Reviews
    Customers Who Bought This Item Also Bought
    View products by: Title | Brand | Category | Price | Popularity | Best Sellers
    View product categories by: Popularity | Our Choices | All-Around Favorites | Title | Product Category | Recommended
    View popular items by: Popularity | Favorites | Title | Price | Part Number | Category
    View Popular Categories by: Popularity | Our Choices | All-Around Favorites | Product Category | Recommended | Title