United States
[Change]
Shopping Cart
Subtotal
=
$0.00
Items
=
0
View Cart
United States
1-800-323-4340
or
Live Chat
Welcome,
log in
or
register
Search
Shop All Products
Balances & Scales
Baths and Circulators
Calibration
Chemicals, Reagents, & Lubricants
Clean Room
Cooling Equipment
Fittings
Flow, Level & Valves
Fluid & Liquid Handling
Furniture & Fume Hoods
Heating Equipment
Industrial Equipment
Lab Equipment
Laboratory Cleaning Supplies
Laboratory Supplies & Consumables
Life Science
Masterflex Peristaltic Pumps
Masterflex Pump Tubing
Material Handling and Storage
Material Testing & Physical Testing
Mixers and Blenders
pH Meters and Water Quality Products
Pressure & Vacuum Instruments
Process Equipment & Instrumentation
Pumps
Recorders & Data Acquisition
Safety Products
Shop by Industry
Spectroscopy and Spectrophotometers
Temperature, Humidity, Moisture and Weather Products
Tubing, and Hose
Shop by
|
Service & Support
|
Technical Resources
|
My Account
Check order status
View Product List
1
of
8 >
Representative photo only
316L Stainless Steel Asme bpe Sanitary Clamp End Cap 20ra Mechanical Finish 1 2 3 4
Click to enlarge
More Product Details
Specifications
Technical Resources
Virtual Catalog (PDFs)
Related technical articles
Service & Support
Answer Box -Q&A posts
Product Reviews
Recommended Products
dummy
Close[X]
SI-30606-29
was succesfully added to your cart.
No Calibration Needed
Yes, Add Calibration
Learn more:
Why calibrate Test Equipment?
Home
>
Fittings
>
Sanitary Fittings, and Clamps
>
ASME-BPE BioPharm sanitary clamp fittings
>
ASME-BPE end caps for clamp fittings
Share
|
316L Stainless Steel Asme-bpe Sanitary Clamp End Cap, 20ra Mechanical Finish, 1/2 - 3/4"
SI-30606-29
$11.00 USD/EACH
In stock.
Qty
:
Add to Cart
Save to shopping list
For your critical applications –fittings meet ASME-BPE 2002 specifications
Interior surface finishes available in both 20Ra Mechanical and 15Ra Electropolished
All fittings are pin-stamped with material grade, heat number(s) and appropriate
ASME-BPE surface finish designation
Mill Test Reports (MTR’s) available on all BPE sanitary fittings
Biopharm products are designed and built to meet the most stringent requirements of ultra-pure processing, including the ASME BPE 2002 specifications for dimensions, surface finishes and tolerances. These fittings are ideal for pharmaceutical, biotechnological and other ultra-pure applications. Biopharm fittings are subjected to rigorous quality control inspections then capped and shrink-wrapped to ensure safe storage and shipping. All Biopharm fittings are constructed of 316L stainless steel.
Roughness Average (Ra) is the arithmetic mean of the peaks and valleys on the surface of the fitting. The lower the Ra reading, the smoother the surface.
Specifications
Product Type
Fittings
Clamp size
0.5 x 0.75
Material
Stainless Steel
Manufacturer number
B16AMP-.50/.75-316L-20/32
Technical Resources
Virtual Catalog (PDFs)
13-14 Catalog
Related technical articles (Roll over for summary)
Sterilizing Fittings: The Critical Factors
Fittings Selection Guide
Show Less...
Service & Support
Still have questions?
Contact Our Experts
for FREE product
application selection
and support.
or call 1-800-323-4340
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here.
Call
1-800-323-4340
Option
9
ext.
6334
See Details
Questions and Answers
Product Reviews
Customers Who Bought This Item Also Bought
Close
Save My Shopping List
Please login in order to use Shopping Lists
Close
Close
Save My Shopping List
You can
view your shopping list
or
continue shopping
View products by:
Title
|
Brand
|
Category
|
Price
|
Popularity
|
Best Sellers
View product categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Title
|
Product Category
|
Recommended
View popular items by:
Popularity
|
Favorites
|
Title
|
Price
|
Part Number
|
Category
View Popular Categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Product Category
|
Recommended
|
Title