Subtotal = $0.00 
Items = 0 
View Cart
United States
1-800-323-4340 or 
Welcome,
log in or register

3M 6000-series Full-face respirators, Medium

EW-40113-12
$168.00 USD/EACH

  • Lightweight and comfortable—ideal for extended wear
  • Exhalation valve directs air downward to prevent fogging of facepiece
Full-facepiece respirators for maximum personal protection. All respirator headstraps are adjustable for a secure fit.
Order respirator facepieces, cartridges, and filters separately according to the demands of your work environment.
Approval: NIOSH
Note: For models 40112-10, -12, and -14, OSHA regulations require gas-proof goggles be worn with half-mask respirators when using formaldehyde.
Product Type Full-face respirators
Color Gray
NIOSH Approval NIOSH
Size Medium
Brand 3M
Manufacturer number 6800

Accessories
Qty/Price
Availability

at $14.00 USD/PACKAGE of 2

In stock.

at $16.25 USD/PACKAGE of 2

In stock.

at $15.50 USD/PACKAGE of 2

In stock.

at $28.00 USD/PACKAGE of 2

In stock.

at $32.00 USD/PACKAGE of 2

In stock.




Technical Resources
Virtual Catalog (PDFs)
  • 13-14 Catalog
  •  
    Service & Support
    Still have questions?
    Contact Our Experts
    for FREE product
    application selection
    and support.
    or call 1-800-323-4340
    Custom Ordering Solutions
    Custom Ordering Solutions
    Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here. Call 1-800-323-4340 Option 9 ext. 6334
    Questions and Answers
    Product Reviews
    Customers Who Bought This Item Also Bought
    View products by: Title | Brand | Category | Price | Popularity | Best Sellers
    View product categories by: Popularity | Our Choices | All-Around Favorites | Title | Product Category | Recommended
    View popular items by: Popularity | Favorites | Title | Price | Part Number | Category
    View Popular Categories by: Popularity | Our Choices | All-Around Favorites | Product Category | Recommended | Title