Subtotal = $0.00 
Items = 0 
View Cart
United States
1-800-323-4340 or 
Welcome,
log in or register

3M Particulate Respirators, N95, Standard Particulate, Box of 20

EW-40111-02
$20.00 USD/BOX of 20

quantity discounts Quantity Discounts
Order Discount Price
[X]
10 10% $18.00/BOX of 20

  • The industry standard for protection against solid and liquid particles
  • Lightweight material for better comfort
  • Adjustable straps for a snug, comfortable fit
Approval: NIOSH
Standard N95 is the economical choice for grinding, sanding, sweeping, bagging, and other work in dusty environments.
Rating N95
Product Type Particulate Respirators
Color White
NIOSH Approval NIOSH
Qty/bx 20
Brand 3M
Manufacturer number 8210





Technical Resources
Virtual Catalog (PDFs)
  • 13-14 Catalog
  •  
    Service & Support
    Still have questions?
    Contact Our Experts
    for FREE product
    application selection
    and support.
    or call 1-800-323-4340
    Custom Ordering Solutions
    Custom Ordering Solutions
    Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here. Call 1-800-323-4340 Option 9 ext. 6334
    Questions and Answers
    Product Reviews
    Customers Who Bought This Item Also Bought
    View products by: Title | Brand | Category | Price | Popularity | Best Sellers
    View product categories by: Popularity | Our Choices | All-Around Favorites | Title | Product Category | Recommended
    View popular items by: Popularity | Favorites | Title | Price | Part Number | Category
    View Popular Categories by: Popularity | Our Choices | All-Around Favorites | Product Category | Recommended | Title