Subtotal = $0.00 
Items = 0 
View Cart
United States
1-800-323-4340 or 
Welcome,
log in or register

3M Three-Panel Particulate respirator, N95, Box of 20

EW-86401-00
$37.00 USD/BOX

quantity discounts Quantity Discounts
Order Discount Price
[X]
10 10% $33.30/BOX

  • Lightweight respirator delivers comfort, convenience, and security
  • Improves visibility, prevents collapsing, and fits many face sizes
  • Individually wrapped to prevent contamination during storage
Low profile makes these respirators ideal for use with other safety equipment. The soft inner panel feels comfortable to the skin. The adjustable nose clip provides a custom fit and a long-lasting, secure seal.
Use in textile, dry chemical, surface preparation, food preparation, woodworking, milling, mineral processing, and other industries that create dusty environments. Respirator can also be used in environments with liquid or non-oil based sprays that do not contain harmful vapors.
Approval: NIOSH
Rating N95
Product Type Particulate Respirators
Color White
NIOSH Approval NIOSH
Qty/bx 20
Brand 3M
Manufacturer number 9210





Technical Resources
Virtual Catalog (PDFs)
  • 13-14 Catalog
  •  
    Service & Support
    Still have questions?
    Contact Our Experts
    for FREE product
    application selection
    and support.
    or call 1-800-323-4340
    Custom Ordering Solutions
    Custom Ordering Solutions
    Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here. Call 1-800-323-4340 Option 9 ext. 6334
    Questions and Answers
    Product Reviews
    View products by: Title | Brand | Category | Price | Popularity | Best Sellers
    View product categories by: Popularity | Our Choices | All-Around Favorites | Title | Product Category | Recommended
    View popular items by: Popularity | Favorites | Title | Price | Part Number | Category
    View Popular Categories by: Popularity | Our Choices | All-Around Favorites | Product Category | Recommended | Title