United States
[Change]
Shopping Cart
Subtotal
=
$0.00
Items
=
0
View Cart
United States
1-800-323-4340
or
Live Chat
Welcome,
log in
or
register
Search
Shop All Products
Balances & Scales
Baths and Circulators
Calibration
Chemicals, Reagents, & Lubricants
Clean Room
Cooling Equipment
Fittings
Flow, Level & Valves
Fluid & Liquid Handling
Furniture & Fume Hoods
Heating Equipment
Industrial Equipment
Lab Equipment
Laboratory Cleaning Supplies
Laboratory Supplies & Consumables
Life Science
Masterflex Peristaltic Pumps
Masterflex Pump Tubing
Material Handling and Storage
Material Testing & Physical Testing
Mixers and Blenders
pH Meters and Water Quality Products
Pressure & Vacuum Instruments
Process Equipment & Instrumentation
Pumps
Recorders & Data Acquisition
Safety Products
Shop by Industry
Spectroscopy and Spectrophotometers
Temperature, Humidity, Moisture and Weather Products
Tubing, and Hose
Shop by
|
Service & Support
|
Technical Resources
|
My Account
Check order status
View Product List
< 2
of
13 >
Representative photo only
47 fiberglass fume hood with no blower 220 VAC 50Hz
Click to enlarge
More Product Details
Specifications
Technical Resources
Virtual Catalog (PDFs)
Related technical articles
Service & Support
Answer Box -Q&A posts
Product Reviews
dummy
Close[X]
EW-33503-05
was succesfully added to your cart.
No Calibration Needed
Yes, Add Calibration
Learn more:
Why calibrate Test Equipment?
Home
>
Furniture & Fume Hoods
>
Fume Hoods
>
Ducted Fume Hoods
>
Labconco 47- and 70-Inch Basic Fume Hoods
Share
|
47" fiberglass fume hood with no blower, 220 VAC/50Hz
EW-33503-05
$4,023.00 USD/EACH
Call 1-888-358-4705 for availability.
Qty
:
Add to Cart
Save to shopping list
Perfect when you need a full-sized ventilated hood to handle small quantities of chemicals
These fume hoods are an ideal alternative to traditional fume hoods when you need a full-sized work area for occasional, light-duty tasks. Models are available with or without a built-in blower, and in standard or explosion-proof configurations. Blowers can be mounted any of three ways—within the hood, above the hood, or on the roof of your building.
All fume hoods include a 3/16" thick tempered safety glass counterbalanced sash and a 10" diameter duct collar. Hoods are constructed of glacier white epoxy-coated 16 gauge steel and include a removable front panel and a 4"L x 4-1/2"W utility pass-through port with molded neoprene cover. Hoods do not include a work surface—order a cabinet with built-in work surface from page 369.
The 110 VAC models are shipped fully assembled; 220 VAC models require assembly. Cord and plug are not included.
Specifications
Product Type
Ducted Fume Hood
Interior dimensions (" W)
47
Exterior Dimensions (" W)
47
Exterior Dimensions (" H)
48
Exterior Dimensions (" D)
25
Blower (hp)
1/3
Power (VAC)
220
Power (Hz)
50
Brand
Labconco
Technical Resources
Virtual Catalog (PDFs)
13-14 Catalog
Related technical articles (Roll over for summary)
Selecting a chemical fume hood: it may be easier than you think!
Importance of Airflow Monitors for Fume Hoods
To B2 or not to B2- a Biological Safety Cabinet Story
Show All...
Show Less...
Service & Support
Still have questions?
Contact Our Experts
for FREE product
application selection
and support.
or call 1-800-323-4340
Leasing Available
Financing is avaliable for many of our products.
See Details
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here.
Call
1-800-323-4340
Option
9
ext.
6334
See Details
Questions and Answers
Product Reviews
Close
Save My Shopping List
Please login in order to use Shopping Lists
Close
Close
Save My Shopping List
You can
view your shopping list
or
continue shopping
View products by:
Title
|
Brand
|
Category
|
Price
|
Popularity
|
Best Sellers
View product categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Title
|
Product Category
|
Recommended
View popular items by:
Popularity
|
Favorites
|
Title
|
Price
|
Part Number
|
Category
View Popular Categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Product Category
|
Recommended
|
Title